Skip to Content
Merck
All Photos(1)

Key Documents

P0234

Sigma-Aldrich

Phorbol 12,13-dinonanoate 20-homovanillate

Synonym(s):

PDNHV

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C47H68O11
Molecular Weight:
809.04
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

solubility

DMSO: soluble
acetone: soluble
chloroform: soluble
ethanol: soluble
methanol: soluble

storage temp.

−20°C

SMILES string

CCCCCCCCC(=O)O[C@@H]1[C@@H](C)[C@@]2(O)C(C=C(COC(=O)Cc3ccc(O)c(OC)c3)C[C@@]4(O)C2C=C(C)C4=O)C5C(C)(C)[C@]15OC(=O)CCCCCCCC

InChI

1S/C47H68O11/c1-8-10-12-14-16-18-20-38(49)57-43-31(4)46(54)34(41-44(5,6)47(41,43)58-39(50)21-19-17-15-13-11-9-2)25-33(28-45(53)37(46)24-30(3)42(45)52)29-56-40(51)27-32-22-23-35(48)36(26-32)55-7/h22-26,31,34,37,41,43,48,53-54H,8-21,27-29H2,1-7H3/t31-,34+,37-,41-,43-,45-,46-,47-/m1/s1

InChI key

FNUXVHZLPOXMCA-WARQNWORSA-N

Biochem/physiol Actions

Resiniferatoxin-type vanilloid phorbol ester with capsaicin-like selectivity for the vanilloid receptor.

Legal Information

Sold in France, Germany, Great Britain and Switzerland under a patent license, restricted to research use only, from Procyon Pharmaceuticals, Inc.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Szallasi et al.
British journal of pharmacology, 128(2), 428-434 (1999-10-08)
1 Although the cloned rat vanilloid receptor VR1 appears to account for both receptor binding and calcium uptake, the identification of vanilloids selective for one or the other response is of importance because these ligands may induce distinct patterns of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service