Skip to Content
Merck

Skip To

P0073

Anti-PERK (N-terminal) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-E2AK3, Anti-EIF2AK3, Anti-HsPEK, Anti-PEK, Anti-PRKR-like endoplasmic reticulum kinase, Anti-Pancreatic eIF2α kinase, Anti-WRS

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
7


biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, rat

technique(s)

western blot: 1:500-1:1,000 using whole cell lysates of HEK-293T expressing PERK

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Eif2ak3(13666)
rat ... Eif2ak3(29702)

General description

PERK (also known as eIF2AK3, PEK and WRS) is a transmembrane kinase that is highly expressed in thepancreas. It resides in the ER and couples stress signals initiated by protein malfolding in the ER lumen to eIF2α phosphorylation and reduces protei
Protein kinase RNA-like endoplasmic reticulum kinase (PERK) is one of the three mammalian UPR (unfolded protein response) transducers; the other two being activating transcription factor 6 (ATF6), and inositol-requiring enzyme 1 (IRE1). It is a highly conserved eIF2a (eukaryotic initiation factor 2 α) kinase.

Immunogen

synthetic peptide corresponding to amino acids 124-139 of mouse PERK, conjugated to KLH. This sequence is identical in rat and mouse.

Application

Anti-PERK (N-terminal) antibody produced in rabbit may be used in immunoblotting.

Biochem/physiol Actions

Anti-PERK (N-terminal) specifically recognizes mouse and rat PERK.
Protein kinase RNA-like endoplasmic reticulum kinase (PERK) is an ER stress sensor which phosphorylates eukaryotic initiation factor 2 α (eIF2α), thereby reducing the translation of most messenger RNAs (mRNAs). This results in reduction of protein load in the ER lumen. In S63del-Charcot-Marie-Tooth 1B neuropathy mice model, the suppression of PERK results in improved myelination. In mice models of frontotemporal dementia, the suppression of translation in a PERK-eIF2α-dependent manner plays a key role in neuronal loss. Administration of GSK2606414, a PERK inhibitor, results in suppression of tau-mediated neurodegeneration. PERK confers protection to heart against pressure overload-induced congestive heart failure. Mutations in this gene are linked with Wolcott-Rallison syndrome (WRS) in humans, which is characterized by permanent neonatal diabetes, exocrine-deficient pancreas, growth retardation, and osteopenia.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Preparation Note

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers is also not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB4301310MABN842R1905
antibody form

IgG fraction of antiserum

antibody form

affinity isolated antibody

antibody form

purified antibody

antibody form

IgG fraction of antiserum

biological source

rabbit

biological source

rabbit

biological source

mouse

biological source

rabbit

conjugate

unconjugated

conjugate

-

conjugate

unconjugated

conjugate

unconjugated

form

buffered aqueous solution

form

buffered aqueous solution

form

-

form

buffered aqueous solution

clone

polyclonal

clone

polyclonal

clone

4F6.1, monoclonal

clone

polyclonal

technique(s)

western blot: 1:500-1:1,000 using whole cell lysates of HEK-293T expressing PERK

technique(s)

immunohistochemistry: 1:50-1:100

technique(s)

immunohistochemistry: suitable (paraffin), western blot: suitable

technique(s)

immunoprecipitation (IP): 5-10 μL using HEK-293T cell lysates, indirect immunofluorescence: 1:100-1:200 using paraformaldehyde-fixed HeLa cells, western blot: 1:500-1:1,000 using HEK-293T cell lystes


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters