Skip to Content
Merck

OGS633

pSF-TAC-NH2-OmpA-FLAG

plasmid vector for molecular cloning

Synonym(s):

cloning vector, expression vector, molecular cloning vector, plasmid, plasmid vector, snapfast vector, vector

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.85
UNSPSC Code:
12352200
Promoter:
Promoter name: Tac
Promoter activity: inducible
Promoter type: bacterial
Origin of replication:
pUC
Bacteria selection:
ampicillin
Peptide cleavage:
EKT

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

pSF-TAC-NH2-OmpA-FLAG, plasmid vector for molecular cloning

recombinant

expressed in E. coli

tag

FLAG® tagged

form

buffered aqueous solution

mol wt

size 5534 bp

bacteria selection

ampicillin

origin of replication

pUC

peptide cleavage

EKT

peptide tag location

N-terminal

promoter

Promoter name: Tac
Promoter activity: inducible
Promoter type: bacterial

secretion signal

OmpA

shipped in

ambient

storage temp.

−20°C

Analysis Note

To view the Certificate of Analysis for this product, please visit www.oxgene.com

Application

Cloning in a gene: This plasmid contains a gene within the main multiple cloning site (NotI-ClaI). Any plasmid that we sell where the gene is this configuration will be located in the exact same position in relation to the start and stop codon of the gene. The only exceptions to this rule are fusions proteins where the fusion gene may be positioned at the front or end of the MCS to allow gene fusion.By positioning all of our genes in the same location it allows them to be transferred between plasmids using the same cloning method and restriction sites regardless of the plasmid being used from our product range. Inserting a new gene into this plasmid should be easily possible using a range of standard restriction enzyme sites that flank the gene currently in the vector.Multiple cloning site notes:In the multiple cloning site there are two important restriction sites called BsgI and BseRI sites. These sites both cut the DNA at the same position and cleave the stop codon of the gene in the multiple cloning site in this plasmid thereby producing a TA overhang. This overhang is compatible with any of our peptide or reporter fusion tag plasmids also cut with either of these enzymes. This allows seamless C-terminal fusions to be made with the gene in this multiple cloning site using a single cloning step from our C-terminal peptide and reporter tag product range. Normally the easiest method is to clone the C-terminal tag from our other plasmid products into this plasmid using BsgI or BseRI and the downstream ClaI restriction site.BseRI and BsgI sites are non-palindromic and cleave a defined number of bases away from their binding sites. This allows them to cut the upstream stop codon in the gene in this plasmid regardless of the gene sequence.

General description

The pSF-TAC-NH2-OmpA-FLAG expression vector is a 5.5 kb E. coli expression vector used for cloning and periplasmic expression of a properly inserted open reading frame as an N-terminal Met-FLAG fusion protein. The FLAG epitope is a small- hydrophilic- 8 amino acid-tag (DYKDDDDK) that enables sensitive detection and high quality protein purification using anti-FLAG products. The N-terminal Met-FLAG fusion protein may be detected using monoclonal anti-FLAG M1 (catalogue number F3040) or M2 (F3165) antibodies. The fusion protein may be purified using anti-FLAG affinity gel (A2220). In this vector, transcription is driven by the IPTG inducible TAC promoter.

Legal Information

Oxford Genetics is a trademark of Oxford Genetics Ltd
FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
Oxford Genetics is a trademark of Oxford Genetics Ltd

Other Notes

Looking for more vector options to move your experiments forward faster? Consider a custom cloning vector designed and built by Oxford Genetics. Find out more at Oxford Genetics - Sigma's partner for cloning and expression vectors for molecular biology and synthetic biology applications.
To view sequence information for this product, please visit the product page

related product

Product No.
Description
Pricing

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service