Skip to Content
Merck

O8014

Org 27569

≥98% (HPLC)

Synonym(s):

5-Chloro-3-ethyl-1H-indole-2-carboxylic acid [2-(4-piperidin-1-ylphenyl)ethyl]amide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C24H28ClN3O
CAS Number:
Molecular Weight:
409.95
UNSPSC Code:
12352200
PubChem Substance ID:
EC Number:
278-169-7
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

color

white

solubility

H2O: <2 mg/mL, DMSO: >5 mg/mL

storage temp.

2-8°C

SMILES string

CCc1c([nH]c2ccc(Cl)cc12)C(=O)NCCc3ccc(cc3)N4CCCCC4

InChI

1S/C24H28ClN3O/c1-2-20-21-16-18(25)8-11-22(21)27-23(20)24(29)26-13-12-17-6-9-19(10-7-17)28-14-4-3-5-15-28/h6-11,16,27H,2-5,12-15H2,1H3,(H,26,29)

InChI key

AHFZDNYNXFMRFQ-UHFFFAOYSA-N

Biochem/physiol Actions

Org 27569 is an allosteric modulator of CB1 cannabinoid receptor.
Org 27569 is an allosteric modulator of CB1 cannabinoid receptor. It significantly increases the binding of CB1 receptor agonist [3H]-CP 55,940 (pKB = 5.67) and causes a significant decrease in specific binding of CB1 receptor inverse agonist SR141716A (Rimonabant; pKB = 5.95); in the reporter gene assay behaves as insurmountable CP 55,940 antagonist; pKB = 7.57 at CB1 receptors in mouse vas deferens


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library