Skip to Content
Merck

O7389

Olprinone hydrochloride

≥98% (HPLC), solid

Synonym(s):

1,2-Dihydro-5-(imidazo[1,2-a]pyridin-6-yl)-6-methyl-2-oxo-3-pyridinecarbonitrile hydrochloride, Loprinone hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C14H10N4O · HCl
CAS Number:
Molecular Weight:
286.72
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
41106305
MDL number:
Assay:
≥98% (HPLC)
Form:
solid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

assay

≥98% (HPLC)

form

solid

color

beige

solubility

DMSO: ≤4 mg/mL, H2O: 6.8 mg/mL

storage temp.

2-8°C

SMILES string

Cl[H].CC1=C(C=C(C#N)C(=O)N1)c2ccc3nccn3c2

InChI

1S/C14H10N4O.ClH/c1-9-12(6-11(7-15)14(19)17-9)10-2-3-13-16-4-5-18(13)8-10;/h2-6,8H,1H3,(H,17,19);1H

InChI key

PWTBMBAQRAOAFF-UHFFFAOYSA-N

Biochem/physiol Actions

Selective phosphodiesterase 3 (PDE3) inhibitor.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emanuela Mazzon et al.
Pharmacological research, 64(1), 68-79 (2011-01-05)
Olprinone is a specific phosphodiesterase (PDE)-III inhibitor, which has been found to have anti-inflammatory effects in addition to its main inotropic and peripheral vasodilatory effects. In the present study we investigated the effects of olprinone (0.2mg/kg, i.p.) on the development
Daniela Mokra et al.
Pulmonary pharmacology & therapeutics, 25(3), 216-222 (2012-03-06)
Since inflammation and oxidation play a key role in the pathophysiology of neonatal meconium aspiration syndrome, various anti-inflammatory drugs have been tested in the treatment. This study evaluated whether the phosphodiesterase (PDE) 3 inhibitor olprinone can alleviate meconium-induced inflammation and
Rosanna Di Paola et al.
European journal of pharmacology, 650(2-3), 612-620 (2010-11-03)
Coronary ischemia and subsequent reperfusion result in deleterious effects, one of the principal ones being vascular and myocardial inflammation. Olprinone hydrochloride, a specific phosphodiesterase III inhibitor, has anti-inflammatory effects in addition to its inotropic and vasodilator effects. The purpose of
Kenya Yamanaka et al.
The Journal of surgical research, 183(1), 391-396 (2012-12-19)
Donor safety is a major concern in living-donor liver transplantation. However, partial grafts do not meet the functional demands of recipients and lead to small-for-size syndrome (SFSS). In a previous study, we showed that olprinone (OLP), a selective phosphodiesterase ІІІ
[Protection from pulmonary apoptosis: a new therapeutic choice for septic acute lung injury].
Ken-ichi Takano et al.
Nihon yakurigaku zasshi. Folia pharmacologica Japonica, 138(4), 146-150 (2011-10-12)

Articles

Cyclic nucleotide phosphodiesterases (PDEs) catalyze the hydrolysis of cAMP and/or cGMP. There are 11 different mammalian PDE families.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Global Trade Item Number

SKUGTIN
O7389-10MG04061832899039
O7389-50MG04061832899046

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service