Skip to Content
Merck
All Photos(1)

Key Documents

O5627

Sigma-Aldrich

N-Oleoyl-D-sphingomyelin semisynthetic from bovine brain sphingomyelin

Synonym(s):

N-(cis-9-Octadecenoyl)-D-sphingosine-1-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H81N2O6P
CAS Number:
Molecular Weight:
729.07
MDL number:
UNSPSC Code:
12352211

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCC\C=C\[C@H](O)[C@@H](COP(O)(=O)OCC[N](C)(C)C)NC(=O)CCCCCCC\C=C/CCCCCCCC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Mattjus et al.
Chemistry and physics of lipids, 71(1), 73-81 (1994-05-06)
In this study we have examined the interaction between cholesterol and phospholipids in monolayers using cholesterol oxidase (Streptomyces cinnamomeus) as a probe. Monolayers containing cholesterol and phospholipids in different molar ratios were exposed to cholesterol oxidase at a lateral surface
Katrin K Halling et al.
Biophysical journal, 95(8), 3861-3871 (2008-07-22)
The lateral organization of lipids and proteins in cell membranes is recognized as an important factor in several cellular processes. Cholesterol is thought to function as a modulator of the lateral segregation of lipids into cholesterol-poor and cholesterol-rich domains. We
T Pawelczyk et al.
Biochimie, 79(12), 741-748 (1998-04-02)
Bovine brain sphingomyelin was separated into different molecular species using a reverse phase column. PLC delta 1 was inhibited by all molecular species of sphingomyelin. The extent of this inhibition was dependent on the hydrophobicity. Based on fatty acid analysis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service