Skip to Content
Merck
All Photos(2)

Key Documents

O1639

Sigma-Aldrich

N-(3-Oxooctanoyl)-DL-homoserine lactone

≥97% (HPLC), white, powder

Synonym(s):

3-Oxo-N-(tetrahydro-2-oxo-3-furanyl)-octanamide, N-(β-Ketooctanoyl)-DL-homoserine lactone

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€117.00

€117.00


Check Cart for Availability


Select a Size

Change View
25 MG
€117.00

About This Item

Empirical Formula (Hill Notation):
C12H19NO4
CAS Number:
Molecular Weight:
241.28
MDL number:
UNSPSC Code:
12352202
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.32

€117.00


Check Cart for Availability

Assay

≥97% (HPLC)

form

powder

color

white

solubility

chloroform: 50 mg/mL, clear, very faintly yellow

shipped in

wet ice

storage temp.

−20°C

SMILES string

CCCCCC(=O)CC(=O)NC1CCOC1=O

InChI

1S/C12H19NO4/c1-2-3-4-5-9(14)8-11(15)13-10-6-7-17-12(10)16/h10H,2-8H2,1H3,(H,13,15)

InChI key

FXCMGCFNLNFLSH-UHFFFAOYSA-N

Biochem/physiol Actions

N-(3-Oxooctanoyl)-DL-homoserine lactone stimulates the tra gene expression. It is an autoinducer and potent antagonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service