Skip to Content
Merck
All Photos(1)

Key Documents

O1266

Sigma-Aldrich

OPC 31260 hydrochloride

≥98% (HPLC)

Synonym(s):

5-(Dimethylamino)-1-[4-(2-methylbenzamido)benzoyl]-2,3,4,5-tetrahydro-1H-benzazepine hydrochloride, Mozavaptan hydrochloride, N-[4-(5-Dimethylamino-2,3,4,5-tetrahydro-1-benzazepine-1-carbonyl)phenyl]-2-methylbenzamide hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€98.90
50 MG
€364.00

€98.90


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€98.90
50 MG
€364.00

About This Item

Empirical Formula (Hill Notation):
C27H29N3O2 · HCl
CAS Number:
Molecular Weight:
464.00
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

€98.90


Estimated to ship on29 May 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

DMSO: ≥15 mg/mL

storage temp.

2-8°C

SMILES string

Cl.CN(C)C1CCCN(C(=O)c2ccc(NC(=O)c3ccccc3C)cc2)c4ccccc14

InChI

1S/C27H29N3O2.ClH/c1-19-9-4-5-10-22(19)26(31)28-21-16-14-20(15-17-21)27(32)30-18-8-13-24(29(2)3)23-11-6-7-12-25(23)30;/h4-7,9-12,14-17,24H,8,13,18H2,1-3H3,(H,28,31);1H

InChI key

MOROBKPIULFQDC-UHFFFAOYSA-N

Biochem/physiol Actions

OPC 31260 is a vasopressin V2 selective antagonist. Arginine vasopressin (AVP) plays an important part in circulatory and water homeostasis. The V2 receptor subtype is important in water retention, cardiac remodeling in congestive heart failure (CHF), and renal hemodynamic alterations, and is considered a target for polysystic kidney disease. OPC-31260 has been shown to inhibit the development of polycystic kidney disease in several animal models.
Vasopressin V2 selective antagonist.

Caution

air-sensitive, hygroscopic, store with desiccant

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li-Hua Xu et al.
The Journal of pharmacology and experimental therapeutics, 373(2), 248-260 (2020-03-14)
It has been identified that arginine vasopressin (AVP), vasopressin receptor 2(V2R), and the aquaporin 2 (AQP2) signaling pathway in the inner ear play important roles in hearing and balance functions through regulating the endolymph equilibrium; however, the contributions of this

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service