Skip to Content
Merck
All Photos(1)

Key Documents

NP4

Sigma-Aldrich

TERGITOL

Type NP-4

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200

description

non-ionic

type

Type NP-4

SMILES string

CCCCCCCCCC1=CC=C(C=C1)OCCO

InChI

1S/C17H28O2/c1-2-3-4-5-6-7-8-9-16-10-12-17(13-11-16)19-15-14-18/h10-13,18H,2-9,14-15H2,1H3

InChI key

KUXGUCNZFCVULO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Polyglycol ether (nonionic) surfactants.

Legal Information

TERGITOL is a trademark of The Dow Chemical Company ("Dow") or an affiliated company of Dow

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service