Skip to Content
Merck
All Photos(1)

Key Documents

N9407

Sigma-Aldrich

Neuropeptide Y Fragment 3-36 human, rat

≥95% (HPLC)

Synonym(s):

NPY 3-36

Sign Into View Organizational & Contract Pricing

Select a Size

250 μG
€196.00

€196.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 μG
€196.00

About This Item

Empirical Formula (Hill Notation):
C175H269N53O54S
CAS Number:
Molecular Weight:
4011.40
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.32

€196.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

powder

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... NPY(4852)

Amino Acid Sequence

Ser-Lys-Pro-Asp-Asn-Pro-Gly-Glu-Asp-Ala-Pro-Ala-Glu-Asp-Met-Ala-Arg-Tyr-Tyr-Ser-Ala-Leu-Arg-His-Tyr-Ile-Asn-Leu-Ile-Thr-Arg-Gln-Arg-Tyr-NH2

General description

Neuropeptide Y (NPY) is a regulatory protein consisting of 36 amino acids. It is ubiquitously expressed in the heart and brain. NPY is a member of pancreatic polypeptide family. The gene encoding this protein is localized on human chromosome 7p15.[1]

Biochem/physiol Actions

Endogenous neuropeptide that selectively binds to Y<SUB>2</SUB> and Y<SUB>5</SUB> receptors.
Neuropeptide Y (NPY) plays a vital role in various processes like feeding behavior, anxiety, epilepsy, circadian rhythm, sleep, pain and drug addiction. It also modulates appetite, body weight and obesity. The protein has potent vasoactive effects on the cardiovascular system and controls central endocrine secretion. NPY is involved in the pathogenesis of atherosclerosis and coronary artery disease.[1][2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genetic studies of neuropeptide Y and neuropeptide Y receptors Y1 and Y5 regions in morbid obesity
C Roche
Diabetologia, 40, 671?675-671?675 (1997)
R Mentlein et al.
Regulatory peptides, 49(2), 133-144 (1993-12-10)
Neuropeptide Y, peptide YY and pancreatic polypeptide share an evolutionary conserved proline-rich N-terminal sequence, a structure generally known to be inert to the attack of common proteinases, but a potential target for specialized proline-specific aminopeptidases. Purified human dipeptidyl peptidase IV
Eitezaz Mahmood et al.
European journal of pharmacology, 882, 173261-173261 (2020-06-14)
Neuropeptide-Y (NPY) leads to angiogenesis and remodeling of the ischemic myocardium. The objective of this study is to assess the therapeutic potential of NPY in a model of acute myocardial ischemia using a nanoparticles delivery system targeted to tissue with
Cloned human neuropeptide Y receptor couples to two different second messenger systems.
Herzog H
Proceedings of the National Academy of Sciences of the USA, 89(13), 5794-5798 (1992)
K N Browning et al.
Neurogastroenterology and motility : the official journal of the European Gastrointestinal Motility Society, 12(1), 33-41 (2000-01-29)
Intracellular recordings were made from myenteric neurones of the guinea-pig descending colon. Neuropeptide Y (NPY) and related pancreatic polypeptides were applied by superfusion and the effects upon the amplitude of fast excitatory synaptic potentials (ESPs) and the ratio of paired

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service