Skip to Content
Merck
All Photos(1)

Key Documents

N7411

Sigma-Aldrich

Neutralization Solution EF

Sign Into View Organizational & Contract Pricing

Select a Size

3.7 L
€97.20

€97.20


Check Cart for Availability


Select a Size

Change View
3.7 L
€97.20

About This Item

UNSPSC Code:
12352200
NACRES:
NA.52

€97.20


Check Cart for Availability

form

liquid

storage temp.

room temp

General description

For sample preparation use in some GenElute kits.

Preparation Note

For usage requirements, please refer to the kit procedure.

Legal Information

GenElute is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service