Skip to Content
Merck
All Photos(1)

Key Documents

N5908

Sigma-Aldrich

N-(NBD-Aminocaproyl)sphinganine

≥98%

Synonym(s):

C6-NBD Sphinganine, N-[6-[(7-Nitro-2-1,3-benzoxadiazol-4-yl)amino]caproyl]-sphinganine

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€55.40

€55.40


Estimated to ship on17 May 2025



Select a Size

Change View
50 μG
€55.40

About This Item

Empirical Formula (Hill Notation):
C30H51N5O6
Molecular Weight:
577.76
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.77

€55.40


Estimated to ship on17 May 2025


Quality Level

Assay

≥98%

form

solid

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCC[C@@H](O)[C@H](CO)NC(=O)CCCCCNc1ccc([N+]([O-])=O)c2nonc12

InChI

1S/C30H51N5O6/c1-2-3-4-5-6-7-8-9-10-11-12-13-15-18-27(37)25(23-36)32-28(38)19-16-14-17-22-31-24-20-21-26(35(39)40)30-29(24)33-41-34-30/h20-21,25,27,31,36-37H,2-19,22-23H2,1H3,(H,32,38)/t25-,27+/m0/s1

InChI key

GEZLBJRDZRUTOE-AHKZPQOWSA-N

General description

Fluorescently labeled fatty acid sphingolipid membrane probe. NBD group excitation 460 nm, emission 534 nm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service