Skip to Content
Merck
All Photos(1)

Documents

N5768

Supelco

(±)-Nicotine-d3 salicylate salt

98 atom % D

Synonym(s):

1-(Methyl-d3)-2-(3-pyridyl)pyrrolidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H17D3N2O3
CAS Number:
Molecular Weight:
303.37
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

isotopic purity

98 atom % D

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental
forensics and toxicology
veterinary

format

neat

mass shift

M+3

SMILES string

OC(=O)c1ccccc1O.[2H]C([2H])([2H])N2CCCC2c3cccnc3

InChI

1S/C10H14N2.C7H6O3/c1-12-7-3-5-10(12)9-4-2-6-11-8-9;8-6-4-2-1-3-5(6)7(9)10/h2,4,6,8,10H,3,5,7H2,1H3;1-4,8H,(H,9,10)/i1D3;

InChI key

AIBWPBUAKCMKNS-NIIDSAIPSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J A Thompson et al.
Journal of chromatography, 231(1), 53-63 (1982-08-13)
Selective extraction and chromatographic techniques have been developed to measure low nanogram quantities of nicotine and cotinine in tissues. Analyses were performed by capillary column gas chromatography with a specific nitrogen-phosphorus detector and by gas chromatography-mass spectrometry. With close structural
Chun Zhang et al.
Oncology reports, 38(6), 3309-3318 (2017-10-19)
Nicotine can stimulate the progression of non-small cell lung cancer (NSCLC) through nicotinic acetylcholine receptors (nAChRs). The persistent proliferation of cancer cells is one of the key effects of nicotinic signaling. The present study aimed to clarify the mechanism of nicotine-induced proliferation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service