Skip to Content
Merck
All Photos(1)

Key Documents

N2404

Sigma-Aldrich

Neurophysin I from bovine pituitary

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.83

form

solid

mol wt

~10 kDa

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Mutations leading to disruptions of the structure and/or conformation of neurophysins are implicated in the pathophysiology of central diabetes insipidus (CDI).

Other Notes

Protein found in vasopressin- and oxytocin-containing neurons in the hypothalamus that is associated with the transport of these hormones to the posterior pituitary.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly purified neurophysin proteins: method of isolation based on their acidic properties.
T K Audhya et al.
Archives of biochemistry and biophysics, 180(1), 130-139 (1977-04-15)
R Kaźmierkiewicz et al.
Acta biochimica Polonica, 44(3), 453-466 (1997-01-01)
This is a review of our recent modeling work aimed at: (i) development and assessment of techniques for reliable refinement of low-resolution protein structures and (ii) using these techniques, at solving specific problems pertinent to neurophysin-bioligand interactions. Neurophysins I and
J K Kim et al.
Proceedings of the Association of American Physicians, 110(5), 380-386 (1998-10-02)
The arginine vasopressin (AVP) precursor gene of mammals contains three exons encoding the principal domains of the polyprotein precursor, including vasopressin (exon A), neurophysin (exon B), and glycopeptide (exon C). The AVP precursor (preprohormone) is processed and transported through the
J J Legros et al.
Hormone research, 45(3-5), 182-186 (1996-01-01)
When they were discovered by Acher and co-workers, neurophysins were thought to act as carriers for the active nonapeptides vasopressin (AVP) and oxytocin (OT) and were then recognized as the inactive fragment of a precursor with a higher molecular weight
F M de Bree et al.
Cellular and molecular neurobiology, 18(2), 173-191 (1998-04-16)
1. In this review the structure-function relationships of the different vasopressin prohormone domains are dated and discussed, with special reference to the neurophysin and glycopeptide domains. 2. The primary structures of the currently known neurophysins and glycopeptide sequences are compared

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service