Skip to Content
Merck
All Photos(3)

Key Documents

N1790

Sigma-Aldrich

Naftifine hydrochloride

Synonym(s):

Naftifine HCL

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H21N · HCl
CAS Number:
Molecular Weight:
323.86
MDL number:
UNSPSC Code:
51102829
PubChem Substance ID:
NACRES:
NA.76
Pricing and availability is not currently available.

Assay

≥99%

form

powder

solubility

ethanol: soluble 50 mg/mL

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria
fungi
yeast

Mode of action

cell wall synthesis | interferes
enzyme | inhibits

storage temp.

2-8°C

SMILES string

Cl.CN(C\C=C\c1ccccc1)Cc2cccc3ccccc23

InChI

1S/C21H21N.ClH/c1-22(16-8-11-18-9-3-2-4-10-18)17-20-14-7-13-19-12-5-6-15-21(19)20;/h2-15H,16-17H2,1H3;1H/b11-8+;

InChI key

OLUNPKFOFGZHRT-YGCVIUNWSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Naftifine is a synthetic, broad spectrum, antifungal agent and allylamine derivative. It is used topically to treat superficial dermatomycoses such as tinea pedis, tinea cruris, and tinea corporis[1].

Biochem/physiol Actions

Allylamine compound that has antifungal and antimycotic activities. Naftifine inhibits fungal squalene epoxidase and prevents ergosterol biosynthesis. Naftifine also has antibiotic activity against Gram-positive and Gram-negative bacteria and has demonstrated anti-inflammatory properties.
Naftifine is an allylamine compound that has antifungal and antimycotic activities. Naftifine inhibits fungal squalene epoxidase and prevents ergosterol biosynthesis. Naftifine also has antibiotic activity against Gram-positive and Gram-negative bacteria. It has demonstrated anti-inflammatory properties such as a reduction in superoxide production and a reduction in polymorphonuclear leukocyte chemotaxis/endothelial adhesion[2]. Naftifine appears to interfere with sterol biosynthesis by inhibiting the squalene 2,3-epoxidase. This inhibition results in decreased amounts of ergosterol and an accumulation of squalene in the cells[1].

Other Notes

Keep container tightly closed in a dry and well-ventilated place.Light sensitive.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Naftifine, a topical allylamine.
Shellie Marks et al.
Journal of drugs in dermatology : JDD, 9(7), 750-750 (2010-08-04)
[The problem of external therapy used in skin mycosis].
G N Tarasenko et al.
Voenno-meditsinskii zhurnal, 325(8), 38-39 (2004-10-09)
Michael H Gold et al.
Skinmed, 9(5), 283-286 (2011-12-15)
Tinea versicolor (TV) is a superficial cutaneous fungal infection characterized by cutaneous pigment changes, pruritus, scaling, and erythema. This open-label, single-center pilot study evaluated the efficacy and safety of naftifine 1% gel applied twice daily for 2 weeks in 10
Lawrence Charles Parish et al.
Journal of drugs in dermatology : JDD, 10(10), 1142-1147 (2011-10-05)
Naftifine HCl 2% cream (NAFT-2%) is a topical allylamine antifungal preparation under development in the U.S. The objective of this randomized, double-blind, vehicle-controlled study was to evaluate the efficacy and safety of a two-week course of once-daily NAFT-2% vs. vehicle
Michael H Gold et al.
Journal of drugs in dermatology : JDD, 11(4), 514-518 (2012-03-29)
Topical antifungal treatment is a mainstay of therapy for Seborrehic Dermatitis (SD). Although the amidazole and ciclopyridine antifungals have been extensively studied, few clinical efficacy data are available for topical allylamine therapy in SD. The objective of this open-label exploratory

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service