Skip to Content
Merck
All Photos(2)

Key Documents

N0893

Sigma-Aldrich

Sodium 1-nonanesulfonate

~98%

Synonym(s):

1-Nonanesulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€253.00
25 G
€685.00

€253.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€253.00
25 G
€685.00

About This Item

Linear Formula:
CH3(CH2)8SO3Na
CAS Number:
Molecular Weight:
230.30
Beilstein:
3738676
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

€253.00


Check Cart for Availability

Request a Bulk Order

description

anionic

Assay

~98%

form

powder

shelf life

limitedshelf life, expiry date on the label

mol wt

230.30 g/mol

technique(s)

protein purification: suitable

mp

>300 °C (lit.)

SMILES string

[Na+].CCCCCCCCCS([O-])(=O)=O

InChI

1S/C9H20O3S.Na/c1-2-3-4-5-6-7-8-9-13(10,11)12;/h2-9H2,1H3,(H,10,11,12);/q;+1/p-1

InChI key

RUYRDULZOKULPK-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Sodium 1-nonanesulfonate has been used in a study to assess the use of para-hydroxybenzoic acid (p-HBA) as an indirect UV detection buffer in capillary zone electrophoresis (CZE). [1] It has also been used in a study to investigate the relationship between circulating lipoprotein-associated phospholipase A2 (Lp-PLA2) levels and coronary plaque volume. [2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Para-hydroxybenzoic acid as an indirect detection buffer in capillary zone electrophoresis
Zhu, P., et al.
Chromatographia, 50, 321-321 (1999)
Tomotaka Dohi et al.
Atherosclerosis, 219(2), 907-912 (2011-10-26)
Lipoprotein-associated phospholipase A2 (Lp-PLA2) is a vascular-specific inflammatory enzyme, of which increases are associated with cardiovascular events. However, the relationship between circulating Lp-PLA2 levels and coronary plaque volume has not been clarified in patients with acute coronary syndrome (ACS). We

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service