Skip to Content
Merck
All Photos(1)

Key Documents

N0164

Sigma-Aldrich

Necrostatin-5

≥98% (HPLC), solid

Synonym(s):

2-[[3,4,5,6,7,8-hexahydro-3-(4-methoxyphenyl)-4-oxo[1]benzothieno[2,3-d]pyrimidin-2-yl]thio]-acetonitrile, 3-p-methoxyphenyl-5,6-tetramethylenothieno[2,3-d]pyrimidin-4-one-2-mercaptoethylcyanide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€85.70

€85.70


Estimated to ship on14 April 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€85.70

About This Item

Empirical Formula (Hill Notation):
C19H17N3O2S2
CAS Number:
Molecular Weight:
383.49
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€85.70


Estimated to ship on14 April 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

solid

color

white to off-white

solubility

DMSO: >10 mg/mL

storage temp.

2-8°C

SMILES string

COc1ccc(cc1)N2C(=O)c3c4CCCCc4sc3N=C2SCC#N

InChI

1S/C19H17N3O2S2/c1-24-13-8-6-12(7-9-13)22-18(23)16-14-4-2-3-5-15(14)26-17(16)21-19(22)25-11-10-20/h6-9H,2-5,11H2,1H3

InChI key

VGONMECBFMCKBS-UHFFFAOYSA-N

Biochem/physiol Actions

Necrostatin-5 is an inhibitor of necroptosis (non-apoptotic cell death pathway) by indirect inhibition of RIP1 kinase.
Necrostatin-5 is an inhibitor of necroptosis (non-apoptotic cell death pathway) by indirect inhibition of RIP1 kinase. Necroptosis is defined as alternative active cell death pathway: Death-Domain Receptor (DRs, e.g. Fas/TNFR) mediated and caspase-inhibitor insensitive with specific morphology (nuclear condensation, organelle swelling, loss of plasma membrane integrity). The necrostatins have been discovered to inhibit this pathway by inhibition of the death domain receptor-associated adaptor kinase RIP1. Three distinct mechanisms appear to be involved: T-loop dependent inhibition by necrostatin-1; partially T-loop independent inhibition by necrostatin-3 and indirect inhibition of RIP1 by necrostatin-5, since necrostatin-5 is a potent inhibitor of immunnoprecipitated RIP1, but does not inhibit recombinant RIP1.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jimstan Periselneris et al.
Immunology, 167(3), 413-427 (2022-07-15)
Published data for the Streptococcus pneumoniae virulence factor Pneumolysin (Ply) show contradictory effects on the host inflammatory response to infection. Ply has been shown to activate the inflammasome, but also can bind to MRC-1 resulting in suppression of dendritic cell

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service