Skip to Content
Merck
All Photos(1)

Key Documents

MTOXRCSUP

Sigma-Aldrich

RPTEC Complete Supplement

sterile-filtered, suitable for cell culture, add to 500 ML of basal medium

Synonym(s):

Basal media supplement for huRPTEC

Sign Into View Organizational & Contract Pricing

Select a Size

30 ML
€123.00

€123.00


Usually ships in 3 weeks.

Request a Bulk Order

Select a Size

Change View
30 ML
€123.00

About This Item

UNSPSC Code:
12352207
NACRES:
NA.81

€123.00


Usually ships in 3 weeks.

Request a Bulk Order

Quality Level

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

storage temp.

−20°C

General description

Sigma has developed a variety of specialized media which are intended to provide optimal performance when utilized with specific cell lines. This product consists of 30 mL of RPTEC complete media supplement which is added to basal media and other components listed below to produce 500 mL of complete media needed for optimal growth, maintenance, and culture of Sigma′s Human Renal Proximal Tubule Epithelial Cells (RPTEC). The final RPTEC complete media was developed specifically for use with Sigma′s human RPTEC control SA7K Clone cells and various knockouts (KO).

Application

RPTEC Complete Supplement has been used in SA7K cell line culture for cell characterization.[1]

Preparation Note

Prepare Complete Medium by supplementing 500 mL of MEMα (M4526) with 6.25 mL of L-Glutamine (G7513), 30 mL of RPTEC Complete Supplement (MTOXRCSUP), 0.3 mL of Gentamicin solution (G1397) and 0.03 mL of Amphotericin B solution (A2942). Complete Medium can be stored at 2–8 °C for up to 1 month. This medium is formulated for use with a 5% CO2 in air atmosphere.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Development and application of human renal proximal tubule epithelial cells for assessment of compound toxicity.
Li S, et al.
Current Chemical Genomics and Translational Medicine, 11, 19-19 (2017)
Marianne K Vormann et al.
The AAPS journal, 20(5), 90-90 (2018-08-16)
Proximal tubules in the kidney play a crucial role in reabsorbing and eliminating substrates from the body into the urine, leading to high local concentrations of xenobiotics. This makes the proximal tubule a major target for drug toxicity that needs

Articles

AIM Biotech's modular platform enables simple to complex 3D cell culture for various research needs.

Human renal proximal tubule epithelial cells (RPTECs) are commonly used to predict human renal drug permeability and to investigate drug efflux. We have generated transporter knockout (KO) cell lines using CompoZr® Zinc Finger Nuclease (ZFN) technology in a proprietary renal proximal tubule epithelial cell line.

Questions

  1. Hello, I was wondering if the FBS in the complete supplement is heat inactivated or not? This would be very helpful for us to understand our recently obtained data. Thank you in advance! With best wishes, Linda

    1 answer
    1. The FBS used to prepare this product has not been heat-inactivated.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service