Skip to Content
Merck
All Photos(1)

Key Documents

MTOX1011

Sigma-Aldrich

PXR Knockout HepaRG Cells

human female liver (Source Disease: Hepatocarcinoma and Hepatitis C)

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
€1,090.00

€1,090.00


Usually ships in 3 weeks.

Request a Bulk Order

Select a Size

Change View
1 VIAL
€1,090.00

About This Item

UNSPSC Code:
12352207
NACRES:
NA.81

€1,090.00


Usually ships in 3 weeks.

Request a Bulk Order

Product Name

PXR Knockout HepaRG Cells, 1 vial

biological source

human female liver (Source Disease: Hepatocarcinoma and Hepatitis C)

Quality Level

form

liquid

OMIM accession no.

storage temp.

−196°C

Gene Information

human ... NR1I2(1337040)

General description

HepaRG is a human hepatoma cell line. The cells possess a pseudodiploid karyotype and have been characterized as an oval ductular bipotent hepatic cell line as they have the ability to differentiate into both biliary and hepatocyte lineages in the presence of DMSO. HepaRG PXR KO cells express the major xenobiotic sensors (CAR and AhR), drug transporters, phase I and II drug metabolizing enzymes as well as key hepatic transcription factors involved in stress response pathways.

Application

See technical bulletin for detailed protocols

Features and Benefits

HepaRG cells are the most metabolically active human hepatocyte cell line developed to date. These cells are suitable for a wide variety of studies for drug metabolism, CYP induction, metabolism-mediated toxicity, transporter, and hepatotoxicity.

  • Sigma′s HepaRG PXR Knockout (KO) allows conclusive identification of CYP3A4 induction through PXR activation.

Zinc finger nucleases (ZFN) mediated Knock-out of NR1I2 (PXR) gene.

  • The frame-shift mutation of NR1I2 gene was confirmed by fragment length analysis and DNA sequencing.
  • Loss of functionality was confirmed by loss of response to inducers of cytochrome P450 activity.

Quality

Tested for Mycoplasma, sterility, post-freeze viability, short terminal repeat (STR) analysis for cell line identification, cytochrome oxidase I (COI) analysis for cell line species confirmation

Legal Information

These products are covered by the License Agreement as described in Exhibit 1 and 2, in the technical bulletin.
HepaRG is a trademark of BioPredic International company

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service