Skip to Content
MTCH68002
MTC Bio Halo Lightweight Single Channel Pipettor
capacity 0.1-2.0 μL
Synonym(s):
pipet, MTC Bio H6800-2, handheld liquid transfer device, liquid transfer device, mechanical pipettor
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.15
UNSPSC Code:
41121500
Packaging:
pack of 1 ea
Feature:
adjustable, manual, no. of channels 1
Manufacturer/tradename:
MTC Bio H6800-2
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
feature
adjustable, manual, no. of channels 1
packaging
pack of 1 ea
manufacturer/tradename
MTC Bio H6800-2
capacity
0.1-2.0 μL
General description
As one of the lightest pipettors ever introduced, the Halo Pipettor dramatically reduces the risk of repetitive stress injury and the general shoulder, arm, and hand fatigue associated with daily pipetting. Reduced actuation forces andan ergonomic design make Halo Pipettors feel balanced and natural in hand. Featuring a four-digit micrometer display for precise volume selection and a click-based adjustment wheel with DriftGuard protection, the Halo Pipettor is perfect for applications that demand exceptional accuracy. With a recessed ejector button, and cushioned, shock-absorbing stop, the ergonomics of tip ejection have been re-imagined to relieve stress on the thumb and joints.
Halo Single Channel Pipettor, lightweight, 20-200µL
Features and Benefits
  • 10-40% lighter than leading competitors
  • 4 Digit micrometer with DriftGuard protection
  • Exclusive 1250µL model for extended length tips
  • User-enabled calibration port
  • Compatible with universal tips
Legal Information
Halo is a trademark of MTC Bio
MTC is a trademark of MTC Bio
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)