Skip to Content
MTCC2530
MTC Bio Five-O Screw Cap MacroTubes®
capacity 5 mL, clear, non-sterile, pkg of 500 ea (caps packed seperately)
Synonym(s):
centrifuge tube, plastic tube, MTC Bio C2530, sample collection tube, sample receptacle, sample tube
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.11
UNSPSC Code:
41121703
Capacity:
5 mL
Material:
clear , conical , polypropylene , screw cap
Sterility:
non-sterile
Packaging:
pkg of 500 ea (caps packed seperately)
Parameter:
25,000 × g speed (max)
Feature:
graduation
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
clear , conical , polypropylene , screw cap
sterility
non-sterile
feature
graduation
packaging
pkg of 500 ea (caps packed seperately)
manufacturer/tradename
MTC Bio C2530
parameter
25,000 × g speed (max)
capacity
5 mL
General description
MTC Bio Five-O 5mL Screw Cap MacroTubes® take all of the conveniences of standard 15mL conical tubes and scales them down for space efficiency and decreased natural resource consumption, they also require about 60% less storage space. Useful from -86°C to 110°C, these tubes are a valuable resource for boiling and for sample preservation at very low temperatures.
5mL Screw-cap MacroTube, non-sterile, w/ screw caps packed separately, 500/cs
Legal Information
MTC is a trademark of MTC Bio
MacroTubes is a registered trademark of MTC Bio
Choose from one of the most recent versions:
Certificates of Analysis (COA)