MT30671558
Mettler Toledo SevenDirect SD20 pH / ORP Meter Pure H2O Kit
electrode, InLab® Pure Pro-ISM
About This Item
Recommended Products
AC/DC input
, universal plug set
220 V / 240 V
feature
electrode InLab® Pure Pro-ISM
packaging
pack of 1 kit
manufacturer/tradename
Mettler-Toledo 30671558
mV measuring range
-2,000.000-2,000.000 mV, accuracy: ±0.1, resolution: 1; 0.1
pH measuring range
-2.000-20.000, accuracy: ±0.002, resolution: 0.1
resolution: 0.001
resolution: 0.01
temperature measuring range
-30-130 °C, resolution: 0.1 °C
General description
Large color touchscreen provides direct measurement and easy data entry. Instructions guide users through the sensor calibration process.
Tracks your Records
The instrument stores data like timestamp, sample and sensor ID with every measurement and prints or exports records automatically.
Tough Enough
Closed connector compartment and replaceable cover keep the instrument safe from dust and spills, even with sensors attached (IP54 in-use).
pH Sensor for Pure Water
InLab Pure-Pro ISM is a pH/temperature sensor designed for accurate and stable pH measurements in pure water. The brige electrolyte, sleeve junction, and low-resistance glass membrane support accurate measurements in ion-poor media.
Electrode Arm for Efficient Operation
The EasyPlace electrode arm makes placing the sensor in the sample or buffer container easy. Dedicated positions for cleaning and storage support correct sensor handling. Vertical and horizontal movements facilitate precise positioning, even with one hand.
Protect Critical Settings
A two-level user management protects the most important settings. Operators retain the flexibility to individually set preferences like language and screen brightness.
Display Adapts to Your Needs
View detailed measurement information on one measurement screen - or focus on the result only and maximize readability with the uFocus mode.
Application
Features and Benefits
- Large color touchscreen provides direct measurement and easy data entry.
- The instrument stores data like timestamp, sample and sensor ID with every measurement and prints or exports records automatically.
- Smart design and cover keeps the instrument safe from dust and spills
- The pH/temperature sensor designed for accurate and stable pH measurements in pure water.
- Electrode arm for efficient operation
- Protect critical settings Protected with two-level user management
- Display adapts to your needs to maximize readability
Components
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service