Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
NACRES:
NA.51
UNSPSC Code:
41105324
Pricing and availability is not currently available.
Quality Level
product line
MISSION®
form
solid
mature sequence
UGGCUCAGUUCAGCAGGAACAG
Sanger mature/minor accession no.
microRNA accession no.
MI0000572
storage temp.
−20°C
1 of 1
This Item | ||
|---|---|---|
| product line MISSION® | product line MISSION® | product line MISSION® |
| microRNA accession no. MI0000572 | microRNA accession no. MI0000142 | microRNA accession no. MI0000713 |
| Sanger mature/minor accession no. | Sanger mature/minor accession no. | Sanger mature/minor accession no. |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
| mature sequence UGGCUCAGUUCAGCAGGAACAG | mature sequence UUCACAGUGGCUAAGUUCUGC | mature sequence CCCAGUGUUCAGACUACCUGUUC |
| form solid | form solid | form solid |
General description
Individual synthetic microRNA inhibitors were designed using a proprietary algorithm, which is based on the work of Haraguchi, T, et al. and in collaboration with Dr. Hideo Iba, University of Tokyo.[1] This algorithm utilizes the tough decoy (TuD) design. miRNA are known to regulate gene expression in a variety of manners, including translational repression, mRNA cleavage and deadenylation.
The MISSION synthetic miRNA Inhibitors are small, double-stranded RNA molecules designed to inhibit a specific mature miRNA. The miRNA inhibitors were designed using the mature miRNA sequence information from miRBase and are 2′-O-methylated RNA duplexes with a miRNA binding site on each strand. Optimal miRNA inhibition is provided after transfection due to the robust secondary structure of the inhibitor.
Two negative controls are available: Arabidopsis thaliana sequence (NCSTUD001) and Caenorhabditis elegans sequence (NCSTUD002)
The MISSION synthetic miRNA Inhibitors are small, double-stranded RNA molecules designed to inhibit a specific mature miRNA. The miRNA inhibitors were designed using the mature miRNA sequence information from miRBase and are 2′-O-methylated RNA duplexes with a miRNA binding site on each strand. Optimal miRNA inhibition is provided after transfection due to the robust secondary structure of the inhibitor.
- Long lasting inhibition at very low dosage
- Excellent resistance to cellular nucleases
- Custom synthesis available for a variety of species
Two negative controls are available: Arabidopsis thaliana sequence (NCSTUD001) and Caenorhabditis elegans sequence (NCSTUD002)
Other Notes
Based on miRBase V19
Legal Information
MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany
signalword
Warning
hcodes
pcodes
Hazard Classifications
STOT RE 2 Inhalation
target_organs
Respiratory Tract
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Global Trade Item Number
| SKU | GTIN |
|---|---|
| MSTUD0102 | 04061836858193 |
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service
