Skip to Content
Merck
All Photos(1)

Key Documents

MSST0010

Sigma-Aldrich

SILuLite MAPK1 Mitogen Activated Protein Kinase 1 human

recombinant, expressed in HEK 293 cells, MS Protein Standard

Synonym(s):

Extracellular signal-regulated kinase 2 (Erk2), Mitogen-activated protein kinase 1 (MAPK1)

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€708.00

€708.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
50 μG
€708.00

About This Item

UNSPSC Code:
23201100
NACRES:
NA.12

€708.00


Estimated to ship on28 May 2025


Request a Bulk Order

recombinant

expressed in HEK 293 cells

Quality Level

tag

His tagged

Assay

≥95% (SDS-PAGE)

form

liquid

mol wt

43.8 kDa

suitability

suitable for mass spectrometry (internal calibrator)

UniProt accession no.

relevant disease(s)

cancer

storage temp.

−20°C

General description

SILuLite MAPK1 is a recombinant human MAPK1 expressed in human 293 cells. It is a monomer of 380 amino acids (including polyhistidine and flag tags), with an apparent molecular weight of 44 kDa. SILuLite MAPK1 is designed to be used as an internal standard for bioanalysis of MAPK1 in mass-spectrometry.

Biochem/physiol Actions

MAPK1 is a cytoplasmic protein that following activation is capable of translocating to the nucleus where it phosphorylates and regulates nuclear proteins (e.g., Elk-1, c-Myc, c-Jun, c-Fos, and C/EBP β). It is a protein serine/threonine kinase that is a member of the extracellular signal-regulated kinases (ERKs) which are activated in response to numerous growth factors and cytokines. Aberrations in the RAS-MAPK complex (of which MAPK1 is a downstream effector) are implicated in several human cancers, and this renders the pathway an attractive therapeutic target.

Sequence

MDYKDDDDKGHHHHHHHHGGQAAAAAAGAGPEMVRGQVFDVGPRYTNLSYIGEGAYGMVCSAYDNVNKVRVAIKKISPFEHQTYCQRTLREIKILLRFRHENIIGINDIIRAPTIEQMKDVYIVQDLMETDLYKLLKTQHLSNDHICYFLYQILRGLKYIHSANVLHRDLKPSNLLLNTTCDLKICDFGLARVADPDHDHTGFLTEYVATRWYRAPEIMLNSKGYTKSIDIWSVGCILAEMLSNRPIFPGKHYLDQLNHILGILGSPSQEDLNCIINLKARNYLLSLPHKNKVPWNRLFPNADSKALDLLDKMLTFNPHKRIEVEQALAHPYLEQYYDPSDEPIAEAPFKFDMELDDLPKEKLKELIFEETARFQPGYRS

Physical form

Supplied as a 0.1 mg/mL solution of 20mM sodium phosphate, pH 8.0, 1M NaCl.

Legal Information

This product is licensed under U.S. Patent No. 7,396,688 and foreign counterparts from E. I. du Pont de Nemours and Company. The purchase of this product conveys to the buyer the nontransferable right to use the purchased amount of the product for research and development only, including services for a third party for consideration. The buyer cannot sell or otherwise transfer this product, its components or materials made using this product or its components to a third party. Information about licenses for excluded uses is available from: E. I. du Pont de Nemours and Company; Attn: Associate Director, Commercial Development; DuPont Experimental Station E268; 200 Powdermill Rd.; Wilmington, DE 19803; 1-877-881-9787 (voice), 1-302-695-1437 (fax), [email protected].
SILu is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service