Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
100 μg | Check Cart for Availability | € 480,00 € 408,00 |
About This Item
NACRES:
NA.25
UNSPSC Code:
12352202
Clone:
-
Species reactivity:
-
Application:
—
Citations:
-
€ 408,00
List Price€ 480,00Save 15%Web-Only Promotion
Check Cart for Availability
form
aqueous solution
shipped in
dry ice
storage temp.
−20°C
General description
SILuMab G is produced by a proprietary process to yield 15N-labeled N-glycans.
Supplied as a frozen solution in phosphate-buffered saline pH 7.5
Application
Package size based on protein content determined by A280 using an extinction coefficient (E0.1%) of 1.4
SILuMab G is ideal for the qualitative and quantitative analysis of glycoprotein glycans, and can be applied to individual glycans or complex mixtures.
Legal Information
This product is licensed under U.S. Patent No. 7,396,688 and foreign counterparts from E. I. du Pont de Nemours and Company. The purchase of this product conveys to the buyer the nontransferable right to use the purchased amount of the product for research and development only, including services for a third party for consideration. The buyer cannot sell or otherwise transfer this product, its components or materials made using this product or its components to a third party. Information about licenses for excluded uses is available from: E. I. du Pont de Nemours and Company; Attn: Associate Director, Commercial Development; DuPont Experimental Station E268; 200 Powdermill Rd.; Wilmington, DE 19803; 1-877-881-9787 (voice), 1-302-695-1437 (fax), [email protected].
SILu is a trademark of Sigma-Aldrich Co. LLC
1 of 1
This Item | |||
|---|---|---|---|
| form aqueous solution | form - | form solid | form solid |
| shipped in dry ice | shipped in wet ice | shipped in wet ice | shipped in wet ice |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
Storage Class
10 - Combustible liquids
wgk
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



