Skip to Content
Merck
All Photos(1)

Key Documents

MSQC10

Sigma-Aldrich

SILuMAb Mouse Stable-Isotope Labeled Monoclonal Antibody

from mouse, recombinant, expressed in CHO cells

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€442.00

€442.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
€442.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.12

€442.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

recombinant

expressed in CHO cells

antibody product type

primary antibodies

Assay

≥90% (SDS-PAGE)

packaging

vial of 100 μg

shipped in

wet ice

storage temp.

−20°C

Related Categories

General description

SILuMAb Mouse is a recombinant, stable isotope-labeled, mouse monoclonal antibody which incorporates [13C6, 15N4]-Arginine and [13C6, 15N2]-Lysine. Expressed in CHO cells, SILuMAb Mouse is designed to be used as a universal internal standard for bioanalysis of monoclonal murine antibodies in animal pharmacokinetics (PK) studies.

SILuMAb Mouse is an IgG1 antibody with a kappa light chain. Because of overlap with common sequences in the Fc region with candidate murine antibodies, SILuMAb Mouse provides universal utility, and thus, eliminates the need to produce candidate-specific internal standards.

Physical form

Each vial of SILuMAb Mouse contains the labeled antibody lyophilized from a solution of phosphate buffered saline. Vial content was determined by measuring A280 and using an extinction coefficient (E0.1%) of 1.4.

Preparation Note

Produced utilizing enriched media containing stable isotope labeled amino acids such as [13C6, 15N4]-Arginine and [13C6, 15N2]-Lysine. Expressed in CHO cells, SILuMAb Mouse is designed to be used as a universal internal standard for bioanalysis of monoclonal murine antibodies in animal pharmacokinetics (PK) studies

Reconstitution

SILuMAb Mouse recovery is maximized when 0.1% formic acid is used for reconstitution of the lyophilized product.
Reconstitution with other solvents may reduce recovery. Do not freeze after reconstitution.

  • Briefly centrifuge the vial at ~10,000 × g to collect the product at the bottom of the vial.
  • Add 500 μL of ultrapure water containing 0.1% formic acid to the vial.
  • Mix the contents by gently inverting the vial a minimum of 5 times.
  • Allow the vial to stand at room temperature for at least 15 minutes and repeat mixing by inversion.

Legal Information

SILu is a trademark of Sigma-Aldrich Co. LLC

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service