Skip to Content
Merck

Skip To

MSG3000

DDC Mass Spect Gold® Serum

Synonym(s):

DDC Serum, Gold Serum for DDC, Mass Spect Gold Serum

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mL
Check Cart for Availability
€ 594,00
1 L
Check Cart for Availability
€ 1.860,00

About This Item

NACRES:
SB.01
UNSPSC Code:
12352204
Form:
liquid
Shipped in:
dry ice

€ 594,00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

Proteomics Grade

form

liquid

shipped in

dry ice

storage temp.

−20°C

General description

DDC Mass Spect Gold® is processed from human plasma using a validated process that reduces the hormone and steroid levels (please see average hormone and steroid values in the Certificate of Analysis). This process also maintains the inherent characteristics of the human plasma resulting in a pool that is consistent with patient samples. The human plasma is collected from donors at FDA licensed facilities. Each donor is screened for donor eligibility according to FDA regulations. Each unit is tested for and found negative for HIV1/2-ab, HBsAg, HCV-ab, HIV-1 (NAT), HCV (NAT) and RPR. The plasma is inspected for conformance to raw material specifications. The plasma is processed according to a batch record. The finished product is analyzed and inspected according to quality control procedures for conformance to specifications. Records from donor unit(s) to final product are maintained according to established procedures. These records include donor records, raw material inspection records, batch records, testing records (baseline through final product), labeling records, final product inspection records and batch release records.

Application

DDC Mass Spect Gold® Serum is used as a zero base or in preparation of controls, calibrators and standards for Immunoassays, GCMS and LCMS applications and Hormone and Steroid mass spectrometry applications.
Common Uses: Base matrix for hormone and steroid calibrators, controls, standards and linearity products.

Other Notes

Each Unit of plasma used in the preparation of this material was tested and found negative or non-reactive for HIV 1/2, HBsAg, HCV, HIV-1 (NAT), HCV (NAT) and RPR by FDA approved methods. Products are intended for research or manufacturing of non-injectable products only.

Legal Information

Mass Spect Gold is a registered trademark of Golden West Biologicals, Inc.

Disclaimer

In-vitro use only. Products are intended for research or manufacturing of non-injectible products only.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MSG4000MSG1000MSG2000
form

liquid

form

liquid

form

liquid

form

liquid

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

grade

Proteomics Grade

grade

Proteomics Grade

grade

Proteomics Grade

grade

Proteomics Grade


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters