Skip to Content
Merck

Skip To

MAS005

Glucose Quick Test Strips

Sufficient for 10 tests

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 kit
Check Cart for Availability
€ 81,00

About This Item

NACRES:
NA.84
UNSPSC Code:
12352106

€ 81,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


input

food(s)
beverage(s)

application(s)

food and beverages

detection method

colorimetric

relevant disease(s)

endocrinological disorders, diabetes

storage temp.

2-8°C

General description

Glucose, which is naturally present in fruits and other plant parts, serves as a primary energy source. Abnormal levels of glucose are associated with metabolic dysfunctions like hypoglycemia, hyperglycemia, and diabetes mellitus. Measuring glucose levels in tissues and body fluids such as blood and urine is commonly done to diagnose glucose-related disorders. Glucose levels are monitored to assess the effectiveness of therapeutics like insulin and sulphonylureas in treating type 2 diabetes. In the food industry, glucose is used both as a natural component and as an additive. It is added to food and beverages not only for its impact on taste, texture, and color but also as a preservative.

Application

The Glucose Quick Test Strips are designed for determining glucose levels in the following applications:
  • Food and Beverage Testing
  • Biological samples such as urine and serum

This kit is suitable for the detection of glucose in a variety of food and beverage samples.

Biochem/physiol Actions

The Glucose Quick Test Strips are based on glucose dehydrogenase catalyzed oxidation of glucose in which the formed NADH reduces a chromogenic reagent. The intensity of the product color is directly proportional to the glucose concentration in the sample.

Features and Benefits

  • Rapid and Versatile: Provides rapid results, offering the choice of using either 20 or 100 μL of sample for semi-quantitative measurement spanning from 0 to 720 mg/L (undiluted) glucose, accommodating diverse testing needs efficiently.
  • Time-Saving: Enables sample treatment and assay completion in under 15 minutes, ensuring efficient testing without compromising accuracy, saving valuable time for users.
  • User-Friendly Design: Simplifies testing with a convenient test strip design, eliminating the need for a plate or cuvette reader, enhancing accessibility and usability for individuals of all skill levels.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MAS004MAS001MAS002
description

Sufficient for 10 tests

description

Sufficient for 10 tests

description

Sufficient for 10 tests

description

Sufficient for 10 tests

detection method

colorimetric

detection method

colorimetric

detection method

colorimetric

detection method

colorimetric

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

input

food(s)
beverage(s)

input

beverage(s)
food(s)

input

food(s)
beverage(s)

input

food(s)
beverage(s)

application(s)

food and beverages

application(s)

food and beverages

application(s)

food and beverages

application(s)

food and beverages

relevant disease(s)

endocrinological disorders, diabetes

relevant disease(s)

-

relevant disease(s)

-

relevant disease(s)

-


Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters