Skip to Content
Merck
All Photos(1)

Key Documents

MAK539

Sigma-Aldrich

DNA Assay Kit

sufficient for 250 fluorometric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84
Pricing and availability is not currently available.

Quality Level

detection method

fluorometric

shipped in

ambient

storage temp.

2-8°C

General description

DNA quantitation is a common practice in molecular biology. Very often DNA is available in minute quantities and the traditional UV 260 nm absorbance method requires microgram quantities for reliable results. Accurate determination of DNA concentration, especially when DNA is present at low concentrations, is crucial for reproducible results in sequencing, cloning, transfection and DNA labeling.

Features and Benefits

  • Linear detection range 2 ng to 40 ng (100 – 2,000 ng/mL) calf thymus DNA.
  • Compatible with high-throughput handling systems.
  • Low interference. RNA, salt (up to 3M NaCl), detergent (< 0.01% SDS) and common DNA extraction buffer do not interfere in the assay.

Suitability

The kit is suitable for quantitative determination of DNA in plasmid DNA, genomic DNA, cDNA, DNA following polymerase chain reaction, and DNA extracted from gel and other matrices.

Principle

The DNA Assay Kit is designed to accurately measure nanogram quantities of plasmid DNA, cDNA, DNA following polymerase chain reaction (PCR), and DNA eluted from gels. The improved method utilizes Hoechst dye that binds specifically with double-stranded DNA. The fluorescence intensity, measured at λEx = 350 nm/λEm = 450 nm, is directly proportional to the DNA concentration in the sample. The optimized formulation substantially reduces interference by substances in the raw samples.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service