Skip to Content
Merck
All Photos(1)

Documents

MAK505

Sigma-Aldrich

Ascorbic Acid Assay Kit

sufficient for 100 colorimetric or fluorometric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

Quality Level

application(s)

food and beverages
pharmaceutical

detection method

colorimetric
fluorometric

shipped in

wet ice

storage temp.

−20°C

Related Categories

General description

Ascorbic acid (the L-enantiomer commonly known as vitamin C) is an important antioxidant found in living organisms and applied as additives in food and other industrial processes. By reacting with reactive oxygen species, it protects the cell from oxidative damage.

Features and Benefits

  • Linear detection range: 6 to 1000 µM for the colorimetric assay and 1 to 100 µM for the fluorometric assay.

Suitability

The kit is suitable for ascorbic acid determination in biological samples such as serum, plasma, urine, saliva, milk, tissue, and cell culture, as well as for studying the effects of drugs on ascorbic acid metabolism.

Principle

The Ascorbic Acid Assay Kit provides a simple, direct and high-throughput assay for measuring ascorbic acid. In this assay, ascorbic acid is oxidized by ascorbate oxidase resulting in the production of H2O2 which reacts with a specific dye to form a pink colored product. The color intensity at 570 nm or fluorescence intensity at λEx = 530 nm/λEm = 585 nm is directly proportional to the ascorbic acid concentration in the sample.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service