Skip to Content
Merck
All Photos(1)

Key Documents

MAK455

Sigma-Aldrich

Whole Blood Hemoglobin (Hb) Kit

Sufficient for 250 colorimetric tests

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€386.00

€386.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
€386.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

€386.00


Check Cart for Availability

Request a Bulk Order

Quality Level

input

blood

detection method

colorimetric

relevant disease(s)

hematological disorder

storage temp.

2-8°C

General description

Hemoglobin (Hb) is made of four globin chains each carrying a heme group. It is carried by red blood cells and transports oxygen from the lungs to the peripheral tissues to maintain the viability of cells. Quantitation of blood hemoglobin has been a key diagnostic parameter for various diseases such as anemia, polycythemia and dehydration.

Application

The Whole Blood Hemoglobin (Hb) Kit may be used for:
  • Hematology Research
  • Drug Discovery

Features and Benefits

  • Fast and Efficient: Get hemoglobin results in just 5 minutes with a quick assay reaction, using only 5 μL of blood, saving time and resources.
  • Safe and Environmentally Friendly: Our assay kit utilizes non-hazardous chemistry, eliminating the need for toxic chemicals like potassium cyanide and hexacyanoferrate(III) used in the traditional Drabkin′s method, ensuring safety and environmental responsibility.
  • Compatibility with High-Throughput Systems: Effortlessly integrate our kit into high-throughput handling systems for streamlined operations, ensuring both efficiency and accuracy in your laboratory processes.

Suitability

The kit is suitable for the quantitative determination of hemoglobin (Hb) in whole blood.

Principle

Simple, direct and automation-ready procedures for measuring hemoglobin concentration are becoming popular in research and drug discovery. The Whole Blood Hemoglobin (Hb) Kit is based on an improved Triton/NaOH method, in which the hemoglobin is converted into a uniform colored end product. The intensity of color, measured at A570 nm, is directly proportional to the hemoglobin concentration in the sample. The optimized formulation substantially reduces interference by substances in the raw samples.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. What is the upper detection limit of MAK455-1KT Whole Blood Hemoglobin (Hb) Kit?

    1 answer
    1. The equivalent Hemoglobin concentration of the Calibrator, which is 12 g/dL, would be utilized as the kit's upper limit of detection.

      Helpful?

  2. Is the use of MAK455 with a diluted whole blood sample suitable?

    1 answer
    1. As long as the diluted samples maintain a hemoglobin content above the detection limit of the kit (0.2g/dL), the kit should function effectively.When the dilution factor of the samples is known, adjusting the calculated concentration by multiplying it with the dilution factor according to the protocol should yield the correct hemoglobin concentration in the samples.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service