Skip to Content
Merck
All Photos(1)

Key Documents

MAK435

Sigma-Aldrich

Acetylcholine Assay Kit

Sufficient for 100 colorimetric or fluorometric tests

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€439.00

€439.00


Estimated to ship on16 April 2025


Request a Bulk Order

Select a Size

Change View
1 KIT
€439.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

€439.00


Estimated to ship on16 April 2025


Request a Bulk Order

Quality Level

input

cell culture(s)
plasma
saliva
urine
milk
serum
tissue

application(s)

pharmaceutical

detection method

colorimetric
fluorometric

relevant disease(s)

cardiovascular diseases, Alzheimer′s disease; neurodegenerative disorders; neurological disorders

storage temp.

−20°C

General description

Acetylcholine is a multifunctional neurochemical with diverse roles in the brain and various organ systems. It acts as a neurotransmitter, facilitating communication between neurons, myocytes, and cells in glandular tissues. The name "acetylcholine" originates from its chemical composition, which is an ester of acetic acid and choline. Tissues that utilize or respond to acetylcholine are termed cholinergic. While it functions as a neurotransmitter throughout the body, acetylcholine is particularly associated with the neuromuscular junction, where it triggers muscle activation by connecting motor neurons in the ventral spinal cord with muscles. Additionally, acetylcholine serves as a neurotransmitter in the autonomic nervous system, enabling communication between preganglionic and postganglionic neurons. It is the final product released by parasympathetic postganglionic neurons and plays vital roles in skeletal muscle movement, regulation of smooth and cardiac muscle, as well as in learning, memory, and mood.

Application

The Acetylcholine Assay Kit may be used for:
  • Neurological Research
  • Drug Discovery
  • Cell culture analysis

Features and Benefits

Convenient and Sensitive Measurement:

  • Utilizes only 20 μL of the sample, providing an accurate determination of acetylcholine.
  • Offers a wide detection range of 10-200 μM (colorimetric) and 0.4-10 μM (fluorometric), ensuring sensitivity across varying concentrations.
Simplified Process:
  • Enjoy a hassle-free procedure requiring the addition of a single working reagent.
  • Complete the assay with just a 30-minute reaction time, saving valuable time and effort.

Suitability

The kit is suitable for the quantitative determination of acetylcholine and evaluation of drug effects on acetylcholine metabolism in serum, plasma, urine, saliva, milk, tissue, and cell culture etc.

Principle

The Acetylcholine Assay Kit provides a simple, direct and high-throughput assay for measuring acetylcholine in biological samples. In this assay, acetylcholine is hydrolyzed by acetylcholinesterase to choline which is oxidized by choline oxidase tobetaine and H2O2. The resulting H2O2 reacts with a specific dye to form a pink colored product. The color intensity at 570 nm or fluorescence intensity at λEx = 530 nm/λEm = 585 nm is directly proportional to the acetylcholine concentration in the sample.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Does the kit compensate for peroxide activity contributed by a compound that could also be an oxidizing agent..

    1 answer
    1. Yes. The kit utilizes a Blank Working Reagent in which Acetylcholinesterase is omitted from the reaction mix. That reagent mix is used to measure endogenous peroxide activity that might be in some test samples.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service