Skip to Content
Merck
All Photos(1)

Documents

MAK422

Sigma-Aldrich

ACE1 Inhibitor Screening Kit (Colorimetric)

sufficient for 100 colorimetric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503

detection method

colorimetric

relevant disease(s)

endocrinological disorders, diabetes; cardiovascular diseases; neurological disorders

storage temp.

−20°C

General description

Angiotensin-I-converting enzyme (ACE) is a peptidyl dipeptidase that catalyzes the conversion of decapeptide angiotensin I to octapeptide angiotensin II, by removing a carboxy-terminal dipeptide. ACE is a key part of the renin angiotensin system that regulates blood pressure. ACE inhibitor inhibits ACE1 enzymatic activity and decreases the production of angiotensin II. As a result, blood vessels are dilated thereby increasing the amount of blood pumped by the heart and lowering the blood pressure. ACE inhibitors are used for treating hypertension, heart failure, stroke etc. Recent research is also focused on the discovery of natural products as ACE1 Inhibitors.

Features and Benefits

ACE1 Inhibitor Screening Kit:
  • Compatible with high-throughput handling systems.

Suitability

The kit is suitable for the screening/characterizing of ACE1 inhibitors.

Principle

The ACE1 Inhibitor Screening Kit can be used to screen potential ACE1 inhibitors. The kit utilizes the ability of an active ACE1 to hydrolyze a synthetic substrate, which results in the decrease in A345 nm. In the presence of Captopril, an ACE1 specific inhibitor, the ACE1 enzymatic activity is greatly reduced and there is no significant decrease in the A345 nm. The assay kit provides a rapid, simple and reliable test for high-throughput screening of ACE1 inhibitors.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service