Skip to Content
Merck
All Photos(2)

Key Documents

MAK184

Sigma-Aldrich

Succinate Colorimetric Assay Kit

Sufficient for 100 Colorimetric tests

Synonym(s):

Succinic Acid Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12161503
NACRES:
NA.84

application(s)

cosmetics
food and beverages

detection method

colorimetric

relevant disease(s)

cardiovascular diseases; immunological diseases

storage temp.

−20°C

General description

Succinate (succinic acid) is a citric acid cycle intermediate and functions in the transport of electrons to the mitochondrial electron transport chain.[1][2] It also functions in innate immune signaling and induces IL-1β production during inflammation. Succinates have been often used for the treatment of hypertension.[3] Succinate measurements can be useful for studying its metabolic, immunological, and therapeutic activities.

Application

Succinate Colorimetric Assay Kit has been used to measure the concentration of succinate.[4][5]

Suitability

Suitable for the detection of succinate in a variety of samples such as cell and tissue culture supernatants and in liquid samples such as wine.

Principle

The Succinate Colorimetric Assay Kit is a highly sensitive assay for determining succinate levels (ranging from 2-10 nmole/well) in a variety of samples. Succinate concentration is determined by a coupled enzyme reaction, which results in a colorimetric (450 nm) product proportional to the succinate present.

Kit Components Only

Product No.
Description

  • Succinate Assay Buffer

  • Succinate Converter

  • Succinate Enzyme Mix

  • Succinate Substrate Mix

  • Succinate Developer

  • Succinate Standard

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

DNA methyltransferase 3b regulates articular cartilage homeostasis by altering metabolism.
Shen J, et al.
JCI insight, 2(12) (2017)
VSIG4 inhibits proinflammatory macrophage activation by reprogramming mitochondrial pyruvate metabolism.
Li J, et al.
Nature Communications, 8(1), 1322-1322 (2017)
Metoprolol succinate extended release/hydrochlorothiazide combination tablets.
Hainer JW and Jennifer S
Vascular Health, 3(3), 279-279 (2007)
Yurong Zhang et al.
Redox biology, 28, 101365-101365 (2019-11-11)
Though succinate accumulation is associated with reactive oxygen species (ROS) production and neuronal injury, which play critical roles in epilepsy, it is unclear whether succinate accumulation contributes to the onset of epilepsy or seizures. We sought to investigate changes in
Sarah L Longnus et al.
Transplantation direct, 8(1), e1265-e1265 (2021-12-23)
Heart transplantation with donation after circulatory death (DCD) has become a real option to increase graft availability. However, given that DCD organs are exposed to the potentially damaging conditions of warm ischemia before procurement, new strategies for graft evaluation are

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service