Skip to Content
Merck
All Photos(2)

Key Documents

MAK173

Sigma-Aldrich

Universal Fluorometric Kinase Assay Kit

sufficient for 250 fluorometric tests

Synonym(s):

Universal Kinase Test Kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€670.00

€670.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
1 KIT
€670.00

About This Item

EC Number:
UNSPSC Code:
12161503
NACRES:
NA.84

€670.00


Estimated to ship on29 May 2025


Request a Bulk Order

usage

sufficient for 250 fluorometric tests

detection method

fluorometric

relevant disease(s)

endocrinological disorders, diabetes; neurological disorders; cardiovascular diseases; cancer

storage temp.

−20°C

General description

Protein kinases are the enzymes that catalyze the transfer of phosphate group from a phosphate donor to an acceptor in a substrate protein. Kinases are of great interest to researchers due to their role in cell signaling and their roles in disease states.

Most of the commercial protein kinase assay kits are based on monitoring either phosphopeptide formation or the ATP depletion. Kinase assays that rely on the detection of phosphopeptides require significant effort to identify an optimized peptide for the specific kinase of interest. The ATP depletion method suffers various interferences due to inhibition or activation of luciferase by compounds found in the biological matrix.

Suitability

Suitable for the determination of kinase activity and for screening and identifying kinase inhibitors

Principle

This Universal Fluorimetric Kinase Assay Kit is based on monitoring ADP formation, which is directly proportional to kinase activity. This enzyme-coupled kit provides a fast, efficient, and homogeneous assay to measure kinase activities. This assay kit is a non-radioactive and non-antibody based method to detect the amount of ADP produced from enzyme reaction without a separation step.

This kit demonstrates high sensitivity (<0.3 μM ADP) and broad ATP tolerance (1-300 μM. This kit can be utilized for determining kinase Michaelis-Menten kinetics and for screening and identifying kinase inhibitors.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service