Skip to Content
Merck
All Photos(3)

Key Documents

MAK078

Sigma-Aldrich

Xanthine Oxidase Activity Assay Kit

sufficient for 100 colorimetric or fluorometric tests

Synonym(s):

XO Activity Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

usage

sufficient for 100 colorimetric or fluorometric tests

detection method

colorimetric
fluorometric

relevant disease(s)

rheumatological diseases; gastrointestinal diseases; cardiovascular diseases

storage temp.

−20°C

Gene Information

human ... XDH(7498)
mouse ... XDH(22436)
rat ... XDH(497811)

General description

Xanthine Oxidase (XO) catalyzes the sequential oxidation of hypoxanthine to xanthine, and xanthine to uric acid and hydrogen peroxide. In humans and other primates, XO controls the final step of purine catabolism and is normally found in the liver and the intestinal mucosa. In rodents, XO is broadly expressed in most tissues. While XO activity is normally very low in blood, liver injury can result in the release of XO into blood. XO may contribute to the pathogenesis of gout and cardiovascular disease, and XO activity or expression may be upregulated in these conditions.

Application

Xanthine Oxidase Activity Assay Kit has been used to measure the xanthine oxidase in serum samples.[1][2]

Suitability

Suitable for the measurement of xanthine oxidase in a variety of samples including tissue, cells and serum

Principle

The Xanthine Oxidase Activity Assay Kit provides a simple and direct procedure for measuring XO activity in a variety of biological samples. XO activity is determined by a coupled enzyme assay, which results in a colorimetric (570 nm)/ fluorometric (λex = 535/λem = 587 nm) product, proportional to the hydrogen peroxide generated. One unit of XO is defined as the amount of enzyme that catalyzes the oxidation of xanthine, yielding 1.0 μmole of uric acid and hydrogen peroxide per minute at 25 °C.

replaced by

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shivangi Rastogi et al.
PLoS pathogens, 17(7), e1009712-e1009712 (2021-07-30)
Mycobacterium tuberculosis (Mtb) has evolved to evade host innate immunity by interfering with macrophage functions. Interleukin-1β (IL-1β) is secreted by macrophages after the activation of the inflammasome complex and is crucial for host defense against Mtb infections. We have previously
Probiotic therapy alleviates hyperuricemia in C57BL/6 mouse model.
Cao T, et al.
Biomedical Research, 28(5) (2017)
Neelam Malik et al.
BMC chemistry, 13(1), 53-53 (2019-08-07)
Hesperitin, a naturally occurring flavonoid was hybridized with phenolic acids to evaluate its potential to inhibit the activity of xanthine oxidase (XO), a key enzyme which catalyses xanthine to uric acid which is found to be associated with gout and
Neelam Malik et al.
Medicinal chemistry (Shariqah (United Arab Emirates)), 16(5), 643-653 (2019-10-05)
Xanthine oxidase (XO; EC 1.17.3.2) has been considered as a potent drug target for the cure and management of pathological conditions prevailing due to high levels of uric acid in the bloodstream. The role of xanthine oxidase has been well
Neelam Malik et al.
BMC chemistry, 13(1), 71-71 (2019-08-07)
Xanthine oxidase is an important enzyme of purine catabolism pathway and has been associated directly in pathogenesis of gout and indirectly in many pathological conditions like cancer, diabetes and metabolic syndrome. In this research rutin, a bioactive flavonoid was explored

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. Does the kit allow the use of cell pellets that have been frozen at -80°C, or do the samples need to be fresh, considering that the cells should be rapidly homogenized with the assay buffer?

    1 answer
    1. The cell pellet can be stored at -80°C with a small amount of PBS, avoiding storage with culture medium. Upon thawing, it's recommended to wash the cells once with PBS and then remove the PBS. Following this step, the assay buffer should be used for extraction according to the kit instructions. It's important to avoid multiple freeze-thaw cycles. However, the supplier does not specify the maximum time frozen cells will remain stable for assay.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service