Skip to Content
Merck
All Photos(2)

Key Documents

MAK061

Sigma-Aldrich

Isocitrate Assay Kit

sufficient for 100 colorimetric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.84

usage

sufficient for 100 colorimetric tests

detection method

colorimetric

storage temp.

−20°C

General description

The new Isocitrate Assay Kit, MAK524, is now available! Isocitrate is a TCA cycle intermediate that is formed from citrate by the activity of the enzyme aconitase. In the TCA cycle, isocitrate is oxidized by isocitrate dehydrogenase (IDH) to α-ketoglutarate with the generation of NAD(P)H. In plants and lower organisms, isocitrate is the branch point from which the glyoxylate shunt operates. Isocitrate is found in substantial concentrations in many fruits and vegetables as well as in foods produced from these raw materials. The Isocitrate Assay Kit provides a simple, sensitive and rapid means of quantifying isocitrate in a variety of samples.

Suitability

Suitable for quantifying isocitrate in a variety of samples including tissue, cells, food and beverages

Principle

Isocitrate concentration is determined by a coupled enzyme assay, which results in a colorometric (450 nm) product, proportional to the isocitrate present. Typical sensitivities of detection for this kit are 1-20 nmoles (0.2 - 5 μg) of isocitrate.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Sigma article discusses tumor cell metabolic pathways, focusing on aerobic glycolysis and mitochondrial activity.

Protocols

We describe here a rapid and sensitive method to separate and measure D-2-OHG and L-2-OHG enantiomers using high-resolution mass spectrometry (HRMS) detection.

Chromatograms

application for HPLC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service