Skip to Content
Merck
All Photos(3)

Key Documents

MAK044

Sigma-Aldrich

Free Fatty Acid Quantitation Kit

sufficient for 100 colorimetric or fluorometric tests

Synonym(s):

Free Fatty Acid Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503

usage

sufficient for 100 colorimetric or fluorometric tests

application(s)

cosmetics
food and beverages

detection method

colorimetric
fluorometric

relevant disease(s)

endocrinological disorders, diabetes; cancer

storage temp.

−20°C

General description

Free fatty acids, also know as nonesterified fatty acids, circulate in the plasma bound to albumin. The majority of free fatty acids are derived from either dietary sources or are mobilized from adipose tissue. Plasma free fatty acids are elevated in many obesity-related disorders and may contribute to insulin resistance in peripheral tissues. Conditions such as sepsis and tumors producing lipoactive homones may also be associated with elevated free fatty acid levels.

Abnormalities in fatty acid metabolism causes fatty liver disease. Accumulation of free fatty acid in myocytes leads to the risk of cardiac dysfunction. Increased fatty acid β-oxidation is implicated in cardiac ischemia.

Application

Free Fatty Acid Quantitation Kit has been used in the following:
  • to measure lipid hydrolysis, by determining the free fatty acid in digested sample.
  • to measure plasma and serum free fatty acids level.
  • to quantify free fatty acids level in liver tissue.

Suitability

Suitable for detecting the long-chain free fatty acids in various biological samples, such as serum, plasma and other body fluids, food, growth media, etc.

Principle

In this kit, the concentration of fatty acids (C8 and longer) is determined by a coupled enzyme assay, which results in a colorimetric (570 nm)/ fluorometric (λex = 535/λem = 587 nm) product, proportional to the fatty acids present.

related product

Product No.
Description
Pricing

replaced by

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Association of Temperament and acute stress responsiveness with Productivity, Feed efficiency, and Methane emissions in Beef cattle: an Observational study
Llonch P, et al.
Frontiers in veterinary science, 3, 43-43 (2016)
Transgenic expression of fatty acid transport protein 1 in the heart causes lipotoxic cardiomyopathy.
Chiu H C, et al.
Circulation Research, 96(2), 225-233 (2005)
Sarmistha Mukherjee et al.
Hepatology (Baltimore, Md.), 65(2), 616-630 (2016-11-04)
The regenerative capacity of the liver is essential for recovery from surgical resection or injuries induced by trauma or toxins. During liver regeneration, the concentration of nicotinamide adenine dinucleotide (NAD) falls, at least in part due to metabolic competition for
Kuifen Ma et al.
Iranian journal of basic medical sciences, 20(11), 1207-1212 (2018-01-05)
Arachidonic Acid/5-lipoxygenase (AA/5-LOX) pathway connects lipid metabolism and proinflammatory cytokine, which are both related to the development and progression of nonalcoholic fatty liver disease (NAFLD). Therefore, the present study was designed to investigate the role of AA/5-LOX pathway in progression
Changes in feed intake during isolation stress in respiration chambers may impact methane emissions assessment.
Llonch P, et al.
Animal Production Science, 58(6), 1011-1016 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service