Skip to Content
Merck
All Photos(1)

Key Documents

M9572

Sigma-Aldrich

MRCKβ (1-473), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

CDC42BPB, KIAA1124

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

102-138 nmol/min·mg

mol wt

~73 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CDC42BPB(9578)

Biochem/physiol Actions

Myotonic Dystrophy Kinase-Related cdc42-binding kinase beta (MRCKb) belongs to the DMPK subfamily. The myotonic dystrophy kinase-related kinases and myotonic dystrophy kinase-related Cdc42 binding kinase (MRCK) are effectors of RhoA and Cdc42, respectively, where they are involved in actin cytoskeletal reorganization and neurite outgrowth. Effects of the repeat expansion on the DMPK gene may be responsible for muscle and heart features of Myotonic Dystrophy.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L T Lam et al.
Human molecular genetics, 9(14), 2167-2173 (2000-08-25)
Myotonic dystrophy (DM) is a multisystemic disorder caused by an inherited CTG repeat expansion which affects three genes encoding the DM protein kinase (DMPK), a homeobox protein Six5 and a protein containing WD repeats. Using a panel of 16 monoclonal
I Tan et al.
The Journal of biological chemistry, 276(24), 21209-21216 (2001-06-16)
The myotonic dystrophy kinase-related kinases RhoA binding kinase and myotonic dystrophy kinase-related Cdc42 binding kinase (MRCK) are effectors of RhoA and Cdc42, respectively, for actin reorganization. Using substrate screening in various tissues, we uncovered two major substrates, p130 and p85

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service