Skip to Content
Merck
All Photos(1)

Key Documents

M9380

Sigma-Aldrich

Met-Ser

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H16N2O4S
CAS Number:
Molecular Weight:
236.29
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (TLC)

form

powder

technique(s)

ligand binding assay: suitable

color

white to off-white

storage temp.

−20°C

SMILES string

O=C(N[C@@H](CO)C(O)=O)[C@H](CCSC)N

InChI

1S/C8H16N2O4S/c1-15-3-2-5(9)7(12)10-6(4-11)8(13)14/h5-6,11H,2-4,9H2,1H3,(H,10,12)(H,13,14)

InChI key

WEDDFMCSUNNZJR-UHFFFAOYSA-N

Biochem/physiol Actions

Met-Ser is a dipeptide.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Venkata N Are et al.
Proteins, 85(12), 2239-2251 (2017-09-21)
Xaa-Pro peptidases (XPP) are dinuclear peptidases of MEROPS M24B family that hydrolyze Xaa-Pro iminopeptide bond with a trans-proline at the second position of the peptide substrate. XPPs specific towards dipeptides are called prolidases while those that prefer longer oligopeptides are
Shane McGoldrick et al.
Biochemical and biophysical research communications, 337(4), 1125-1132 (2005-10-19)
The TEF4 gene of the non-saccharomyces yeast Yarrowia lipolytica encodes an EF1Bgamma protein with structural similarity to the glutathione transferases (GSTs). This 1203bp gene was cloned, over-expressed in Escherichia coli, and the recombinant protein characterized. DNA sequencing of the cloned

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service