Skip to Content
Merck
All Photos(1)

Key Documents

M9309

Sigma-Aldrich

McCoy′s 5A Medium

With ʟ-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

5A basal medium

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
€54.10
6 X 500 ML
€250.00

€54.10


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
500 ML
€54.10
6 X 500 ML
€250.00

About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.71

€54.10


Estimated to ship on22 May 2025


Request a Bulk Order

Product Name

McCoy′s 5A Medium, Modified, with L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

components

NaHCO3: 2.2 g/L
L-glutamine: 0.21 g/L
phenol red: 0.011 g/L
glucose: 3.0 g/L (Dextro)

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

The amino acid requirements for in vitro cultivation of Novikoff hepatoma cells were reported by McCoy and colleagues. These experiments were carried out using Basal Medium 5A, which was later modified to create McCoy′s 5A medium.

Application

McCoy′s 5A Medium has been used:
  • to culture HCT-116?cells[1]
  • to culture clear cell renal cell carcinoma (ccRCC) cells Caki-1[2]
  • to maintain and culture human bone osteosarcoma Saos-2 cell line for in vitro cell studies[3]

Biochem/physiol Actions

McCoy′s 5A Medium has been originally developed to support liver tumor cells by modification of the amino acids found in BME. This formulation has also been used to support the growth of primary cultures of bone marrow, skin, gingiva, kidney, omentum, adrenals, lung, spleen, rat embryos, and other cell types.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aleksandr Ianevski et al.
Cancers, 12(6) (2020-07-08)
The evidence that pan-Bcl-2 or Bcl-xL-specific inhibitors prematurely kill virus-infected or RNA/DNA-transfected cells provides rationale for investigating these apoptotic inducers further. We hypothesized that not only invasive RNA or DNA (biological factors) but also DNA/RNA-damaging chemical or physical factors could
Alison Roth et al.
Nature communications, 9(1), 1837-1837 (2018-05-11)
Malaria liver stages represent an ideal therapeutic target with a bottleneck in parasite load and reduced clinical symptoms; however, current in vitro pre-erythrocytic (PE) models for Plasmodium vivax and P. falciparum lack the efficiency necessary for rapid identification and effective
Xiang-Ping Liu et al.
Journal of experimental & clinical cancer research : CR, 28, 52-52 (2009-04-21)
RhoA and RhoC are deregulated by over expression in many human tumors, including colorectal cancer. Some reports show that they play a pivotal role in the carcinogenesis, tumor development and infiltration metastasis. In this study, for the first time we
Nengquan Sheng et al.
Cancer medicine, 6(6), 1331-1340 (2017-04-26)
MicroRNA-145 (miR-145), as a tumor-suppressive miRNA, has been demonstrated down-regulated in colorectal cancer (CRC) cells, and could inhibit CRC cells growth. However, the molecular pathway in which miR-145 modulates CRC malignant transformation has not been fully revealed. Here, we reported
Qiao Zhang et al.
International journal of oncology, 57(1), 197-212 (2020-04-23)
Glucose‑6‑phosphate dehydrogenase (G6PD) is crucial rate‑limiting enzyme of the pentose phosphate pathway (PPP). G6PD dysregulation has been reported in various types of human cancer, and the role of G6PD in cancer progression was demonstrated in numerous studies. A previous study

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service