Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
1 ea | Check Cart for Availability | € 1.530,00 |
About This Item
NACRES:
NA.56
UNSPSC Code:
23151817
€ 1.530,00
Check Cart for Availability
Application
Mimetic columns are used in affinity chromatography and protein chromatography. Mimetic columns have been used in studies to develop anti-inflammatory therapeutics.
1 of 1
This Item | |||
|---|---|---|---|
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
| description 10 ml | description 10 ml | description - | description 10 ml |
| form suspension | form suspension | form suspension | form suspension |
| shipped in wet ice | shipped in wet ice | shipped in wet ice | shipped in wet ice |
signalword
Warning
hcodes
Hazard Classifications
Flam. Liq. 3
Storage Class
3 - Flammable liquids
wgk
WGK 1
flash_point_f
95.0 °F - closed cup
flash_point_c
35 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




