Skip to Content
Merck

M8690

Methyl pristanate

≥99% (GC), liquid

Synonym(s):

Methyl 2,6,10,14-Tetramethylpentadecanoate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H40O2
CAS Number:
Molecular Weight:
312.53
UNSPSC Code:
12352200
PubChem Substance ID:
EC Number:
200-659-6
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥99% (GC)

form

liquid

storage temp.

−20°C

SMILES string

COC(=O)C(C)CCCC(C)CCCC(C)CCCC(C)C

Biochem/physiol Actions

Methyl branched fatty acid intermediate of pristanic acid, which is involved in peroxisomal disorders.


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

51.8 °F - closed cup

flash_point_c

11 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

67-56-1

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library