Skip to Content
Merck
All Photos(1)

Key Documents

M8255

Sigma-Aldrich

Myristoyl chloride

≥99%

Synonym(s):

Tetradecanoyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)12COCl
CAS Number:
Molecular Weight:
246.82
Beilstein:
636924
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥99%

form

liquid

refractive index

n20/D 1.449 (lit.)

bp

250 °C/100 mmHg (lit.)

mp

−1 °C (lit.)

density

0.908 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCC(Cl)=O

InChI

1S/C14H27ClO/c1-2-3-4-5-6-7-8-9-10-11-12-13-14(15)16/h2-13H2,1H3

InChI key

LPWCRLGKYWVLHQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Covalent attachment of myristate is a common occurrence on a wide variety of viral and cellular proteins and plays a role in promoting membrane binding.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

J Goldberg
Cell, 95(2), 237-248 (1998-10-28)
Ras-related GTPases are positively regulated by guanine nucleotide exchange factors (GEFs) that promote the exchange of GDP for GTP. The crystal structure of the Sec7 domain GEF bound to nucleotide-free ARF1 GTPase has been determined at 2.8 A resolution and
M D Resh
Biochimica et biophysica acta, 1451(1), 1-16 (1999-08-14)
Covalent attachment of myristate and/or palmitate occurs on a wide variety of viral and cellular proteins. This review will highlight the latest advances in our understanding of the enzymology of N-myristoylation and palmitoylation as well as the functional consequences of
Janclei P Coutinho et al.
Talanta, 134, 256-263 (2015-01-27)
In this work a multivariate statistical tool (Derringer and Suich optimization) was proposed for the separation of seventeen capsinoids (natural and synthetic) using the UHPLC-DAD chromatography. Capsinoids were analyzed at 280 nm. The variables optimized were the mobile phase (water

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service