Skip to Content
Merck
All Photos(1)

Key Documents

M8194

Sigma-Aldrich

Anti-MAFF (C-Terminal) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-v-maf Avian Musculoaponeurotic Fibrosarcoma Oncogene Family, ProteinF

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
€641.00

€641.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
200 μL
€641.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€641.00


Estimated to ship on24 May 2025


Request a Bulk Order

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 18 kDa

species reactivity

human

technique(s)

immunoprecipitation (IP): 2-4 μL using A431 cell line
microarray: suitable
western blot: 1:2,000-1:4,000 using whole extracts of the A431 cell line

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MAFF(23764)

Related Categories

General description

The MAFF gene is located on the human chromosome at 22q13.1. The encoded protein is a small MAF protein and is expressed in the nucleus.
v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog F (MAFF) is a transcription factor which does not have a transactivation domain but possesses a leucine-zipper.

Specificity

Anti-MAFF (C-terminal) specifically recognizes MAFF.

Immunogen

synthetic peptide corresponding to amino acids 147-163 of human MAFF (isoform a). The immunizing peptide does not cross-react with other members of the family.

Application

Anti-MAFF (c-terminal) antibody produced in rabbit has been used in:
  • western blotting[1]
  • chromatin immunoprecipitation (chip)[1]
  • co-immunoprecipitation[1]
  • immunoprecipitation

Biochem/physiol Actions

The small MAF proteins are associated with many physiological processes including development, differentiation, hematopoiesis and stress response. Interestingly, these proteins regulate the stress response via different mechanisms. The MAFF gene is down-regulated in diabetes mellitus 1 patient. This gene is underexpressed in acute lymphoblastic lymphoma (ALL) patients.
v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog F (MAFF) has been shown to interact with other transcription factors and it associates with certain regulatory sequences in the gene encoding the oxytocin receptor.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, and 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhenyi Qiao et al.
Frontiers in nutrition, 9, 799053-799053 (2022-04-05)
Metabolites of probiotics that are beneficial to human health have been isolated from the intestinal tract and natural dairy products. However, many studies on probiotics and prebiotics are limited to the observation of human cohorts and animal phenotypes. The molecular
Meenakshi B Kannan et al.
Biochimica et biophysica acta, 1823(10), 1841-1846 (2012-06-23)
The small MAFs, MAFF, MAFG and MAFK have emerged as crucial regulators of mammalian gene expression. Previous studies have linked small MAF function, by virtue of their heterodimerization with the Cap 'n' Collar (CNC) family of transcription factors, to the
Yaeko Takagi et al.
Biochemical and biophysical research communications, 320(1), 62-69 (2004-06-23)
Small Maf proteins play critical roles on morphogenesis and homeostasis through associating with CNC proteins. To date, three small Maf proteins, MafF, MafG, and MafK, have been reported in vertebrates, which share redundant functions. In this study, we tried to
Julie A Moran et al.
The Biochemical journal, 361(Pt 2), 371-377 (2002-01-05)
The three small Maf proteins, MafF, MafG and MafK, have been implicated in a number of physiological processes, including development, differentiation, haematopoiesis and stress response. Here we report the constitutive expression of mafF, mafG and mafK in six human cell
Eui Jung Moon et al.
Nature communications, 12(1), 4308-4308 (2021-07-16)
Hypoxia plays a critical role in tumor progression including invasion and metastasis. To determine critical genes regulated by hypoxia that promote invasion and metastasis, we screen fifty hypoxia inducible genes for their effects on invasion. In this study, we identify

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service