Skip to Content
Merck
All Photos(1)

Key Documents

M8072

Sigma-Aldrich

Anti-Mucolipin-1 antibody, Mouse monoclonal

clone MLN128, purified from hybridoma cell culture

Synonym(s):

Anti-MCOLN1, Anti-ML4, Anti-MLIV, Anti-MST080, Anti-MSTP080, Anti-Mucolipidin, Anti-TRP-ML1, Anti-TRPM-L1, Anti-TRPML1, Anti-Transient receptor potential cation channel, mucolipin subfamily, member 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

MLN128, monoclonal

form

buffered aqueous solution

mol wt

antigen ~110 kDa (additional bands may be observed)

species reactivity

human

packaging

antibody small pack of 25 μL

concentration

~2.0 mg/mL

technique(s)

immunocytochemistry: suitable
indirect ELISA: suitable
western blot: 4-8 μg/mL using membrane fraction of HEK-293T expressing human mucolipin-1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MCOLN1(57192)

General description

Monoclonal Anti-Mucolipin-1 (mouse IgG1 isotype) is derived from the hybridoma MLN128 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a recombinant fusion protein corresponding to amino acid 1.Mucolipin-1 is also termed TRP-ML1, MLN1, ML1 mucolipidin. MLN1 shares significant sequence homology with the TRP superfamily of cation channels.
Mucolipin-1 (MCOLN1) is a member of transient receptor potential (TRP) protein family. It is a cation channel present on endosomes and lysosomes.

Application

Monoclonal Anti-Mucolipin-1 has been used in:
  • enzyme linked immunosorbent assay (ELISA)
  • immunoblotting
  • immunocytochemistry.

Biochem/physiol Actions

Mucolipin-1 (MCOLN1) is involved in the regulation of lysosomal trafficking. It aids in the transport of Ca2+ into the cytosol from the lumen, in response to the changing levels of phosphatidylinositol-3, 5-bisphosphate. Mutations in the gene encoding MCOLN1 have been shown to be associated with mucolipidosis type IV.
Mutations in the MCOLN1 gene is implicated in Mucolipidosis type IV (MLIV) is an autosomal recessive, neurodegenerative disorder. Rather, MLIV pathophysiology has been linked to deficiency in membrane trafficking, and organelle dynamics in the late endocytic pathway. Specifically, MLIV cells have been shown to accumulate autophagosomes, due to increased de novo autophagosome formation and due to delayed fusion of autophagosomes with late endosomes/lysosomes.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Posttranslational cleavage and adaptor protein complex-dependent trafficking of mucolipin-1
Miedel MT, et al.
The Journal of Biological Chemistry, 281(18), 12751-12759 (2006)
Autophagic dysfunction in mucolipidosis type IV patients
Vergarajauregui S, et al.
Human Molecular Genetics, 17(17), 2723-2737 (2008)
Helen Waller-Evans et al.
Biochemical Society transactions, 43(3), 442-446 (2015-05-27)
TRPML1 is a ubiquitously expressed cation channel found on lysosomes and late endosomes. Mutations in TRPML1 cause mucolipidosis type IV and it has been implicated in Alzheimer's disease and HIV. However, the mechanisms by which TRPML1 activity is regulated are not
Zhenxing Liu et al.
Biochemistry and cell biology = Biochimie et biologie cellulaire, 92(4), 279-286 (2014-06-25)
Lysosomotropic amines cause serious side effects such as cytoplasmic vacuolation and cell death. TRPML1 (also known as mucolipin1), a member of the transient receptor potential (TRP) protein family, may regulate fusion/fission of vesicles along the endocytic pathway and some aspects
Wuyang Wang et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(11), E1373-E1381 (2015-03-04)
Upon nutrient starvation, autophagy digests unwanted cellular components to generate catabolites that are required for housekeeping biosynthesis processes. A complete execution of autophagy demands an enhancement in lysosome function and biogenesis to match the increase in autophagosome formation. Here, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service