Skip to Content
Merck
All Photos(2)

Key Documents

M7890

Sigma-Aldrich

1-Monopalmitoleoyl-rac-glycerol

≥99%

Synonym(s):

1-Mono(cis-9-hexadecenoyl)-rac-glycerol, Glyceryl 9-hexadecenoate, Monopalmitolein

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H36O4
CAS Number:
Molecular Weight:
328.49
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

Assay

≥99%

form

low-melting solid

functional group

ester

lipid type

neutral glycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCC/C=C\CCCCCCCC(OCC(O)CO)=O

InChI

1S/C19H36O4/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-19(22)23-17-18(21)16-20/h7-8,18,20-21H,2-6,9-17H2,1H3/b8-7-

InChI key

KVYUBFKSKZWZSV-FPLPWBNLSA-N

Biochem/physiol Actions

Monopalmitolein is used in comparative studies of the antimicrobial activity of monoglycerides such as monocaprin, and monlaurin.

Packaging

Sealed ampule.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J C McLay et al.
International journal of food microbiology, 73(1), 1-9 (2002-03-09)
The lactoperoxidase system (LPS) and monolaurin (ML) are potential natural antimicrobial agents for use in foods. The LPS is considered to have greatest activity against Gram-negative bacteria while ML is usually considered to have greatest activity against Gram-positive bacteria. An
G Bergsson et al.
Antimicrobial agents and chemotherapy, 42(9), 2290-2294 (1998-09-16)
The antichlamydial effects of several fatty acids and monoglycerides were studied by incubating Chlamydia trachomatis bacteria with equal volumes of lipid solutions for 10 min and measuring the reduction in infectivity titer compared with that in a control solution without
Alexandru Zabara et al.
Nanoscale, 9(2), 754-763 (2016-12-16)
The structural changes occurring at the nanoscale level within the lipid bilayer and driving the in-meso formation of large well-diffracting membrane protein crystals have been uniquely characterized for a model membrane protein, intimin. Importantly, the order to order transitions taking
Amanda A Frick et al.
The Analyst, 144(11), 3590-3600 (2019-05-09)
More detailed fundamental information is required about latent fingermark composition in order to better understand fingermark properties and their impact on detection efficiency, and the physical and chemical changes that occur with time following deposition. The composition of the glyceride
Jayesh A Kulkarni et al.
Nanoscale, 9(36), 13600-13609 (2017-09-07)
A straightforward "bottom-up" synthesis is described for efficient entrapment of inorganic hydrophobic nanoparticles (HNPs) consisting of iron oxide, gold, or quantum dots within the hydrophobic core of lipid nanoparticles (LNPs). These LNPs consist of hydrophobic "core" lipids such as triolein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service