Skip to Content
Merck
All Photos(1)

Documents

M7273

Sigma-Aldrich

Anti-Microtubule-Associated Proteins (MAPs) antibody produced in rabbit

whole antiserum

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

contains

15 mM sodium azide

species reactivity

mammals

technique(s)

western blot: 1:200

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Microtubule-associated protein 2 (MAP2) is a neuronal protein. It belongs to the MAP2/Tau family. This gene is located to human chromosome 2q34-q35. MAP2 is present in neurons. MAP2 is the major microtubule-associated protein of brain tissue. There are two major classes of heat stable MAPs; two are similarly sized with apparent molecular weights of 280kDa (MAP2a and MAP2b) and the third with a lower molecular weight of 70kDa (MAP2c).

Specificity

Reacts with the two major classes of heat stable MAPs (MAP2 and τ). Does not react with MAP1 or tubulin.

Immunogen

heat stable MAPs prepared from bovine brain.

Application

Anti-Microtubule-Associated Proteins (MAPs) antibody produced in rabbit has been used in:
  • immunofluorescent labelling
  • immunohistochemistry
  • immunoprecipitation
  • immunofluorescence staining
  • western blotting

Biochem/physiol Actions

Microtubule-associated protein 2 (MAP2) interacts with microtubules to maintain the structure of dendrites. It plays a major role in supporting the actin cytoskeleton in spines, binding and nucleating filamentous actin (f-actin) to modulate spine morphology. Microtubules function as structural and mobility elements in mitosis, intracellular transport, flagellar movement and in the cytoskeleton.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Increased Phosphorylation of Ca/Calmodulin-dependent Protein Kinase II and Its Endogenous Substrates in the Induction of Long Term Potentiation
Fukunaga K, et al.
Test, 270(11), 6119-6124 (1995)
Expression of multiple functional chemokine receptors and monocyte chemoattractant protein-1 in human neurons
Coughlan CM, et al.
Neuroscience, 97(3), 591-600 (2000)
Suzana Angelica Silva Lustosa et al.
TheScientificWorldJournal, 2014, 102541-102541 (2014-04-17)
Colorectal cancer dissemination depends on extracellular matrix genes related to remodeling and degradation of the matrix structure. This investigation intended to evaluate the association between FN-1, ITGA-3, ITGB-5, MMP-2, and MMP-9 gene and protein expression levels in tumor tissue with
N Noshita et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 21(5), 557-567 (2001-05-03)
Release of cytochrome c from mitochondria to cytosol is a critical step in the mitochondrial-dependent signaling pathways of apoptosis. The authors have reported that manganese superoxide dismutase (Mn-SOD) attenuated cytochrome c release and apoptotic cell death after focal cerebral ischemia
K L Loveland et al.
Biology of reproduction, 54(4), 896-904 (1996-04-01)
The testis is one of the most abundant sources of microtubule networks. These networks include mitotic and meiotic spindles, the spermatid manchette and axoneme, and the Sertoli cell cytoskeleton. Microtubules are composed of alpha- and beta-tubulin subunits that are polymerized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service