Skip to Content
Merck
All Photos(1)

Key Documents

M6682

Sigma-Aldrich

Methoprene acid

≥98% (TLC)

Synonym(s):

11-Methoxy-3,7,11-trimethyl-2E,4E-dodecadienoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H28O3
CAS Number:
Molecular Weight:
268.39
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (TLC)

storage temp.

−20°C

SMILES string

COC(C)(C)CCCC(C)C\C=C\C(C)=C\C(O)=O

InChI

1S/C16H28O3/c1-13(10-7-11-16(3,4)19-5)8-6-9-14(2)12-15(17)18/h6,9,12-13H,7-8,10-11H2,1-5H3,(H,17,18)/b9-6+,14-12+

InChI key

MNYBEULOKRVZKY-TZOAMJEDSA-N

General description

Retinoic acid receptor (RXR) agonist used in gene regulation research. Analog of juvenile hormone, an insect growth hormone.

Biochem/physiol Actions

Demethylation, oxidative cleavage, hydrolysis, photodegradation and degradation of methoprene by microorganism results in methoprene acid and other metabolites. Methoprene acid is an insect growth regulator, and is known to control ecdysteroid signaling during development and metamorphism. It is thus used to control larval growth in insects such as mosquitoes, cigarette beetles, fleas and fire ants.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M A Harmon et al.
Proceedings of the National Academy of Sciences of the United States of America, 92(13), 6157-6160 (1995-06-20)
We report that methoprene and its derivatives can stimulate gene transcription in vertebrates by acting through the retinoic acid-responsive transcription factors, the retinoid X receptors (RXRs). Methoprene is an insect growth regulator in domestic and agricultural use as a pesticide.
Juvenile Hormones: Advances in Research and Application (2012)
Rintaro Suzuki et al.
Scientific reports, 1, 133-133 (2012-02-23)
Juvenile hormone (JH) plays crucial roles in many aspects of the insect life. All the JH actions are initiated by transport of JH in the hemolymph as a complex with JH-binding protein (JHBP) to target tissues. Here, we report structural
Occurrence and fate of methoprene compounds in urban areas of Southern Ontario, Canada
Struger J, et al.
Bulletin of Environmental Contamination and Toxicology, 79(2), 168-171 (2007)
Daniel Jahn et al.
Biochimica et biophysica acta, 1859(2), 381-392 (2016-01-03)
Fibroblast growth factor 19 (FGF19) is a gut-derived hormone that controls bile acid (BA), carbohydrate and lipid metabolism. Whereas strong evidence supports a key role of BAs and farnesoid X receptor (FXR) for the control of FGF19 expression, information on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service