Skip to Content
Merck
All Photos(1)

Key Documents

M6562

Greiner 96 well 1 x 8 strip plates

high binding

Synonym(s):

96 multiwell plates, 96 well microplates, 96 well microtiter plates, 96 well strip plates

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
€647.00

€647.00


Estimated to ship on16 May 2025



Select a Size

Change View
100 EA
€647.00

About This Item

UNSPSC Code:
41122107
NACRES:
NB.22
material:
clear polystyrene (plate strips)
conical bottom clear polystyrene wells
size:
96 wells
sterility:
non-sterile
binding type:
high binding surface
feature:
lid: no

€647.00


Estimated to ship on16 May 2025


material

clear polystyrene (plate strips)
conical bottom clear polystyrene wells

sterility

non-sterile

feature

lid: no

packaging

case of 100

manufacturer/tradename

Greiner 705071

maximum volume

346 μL

size

96 wells

working volume

300 μL

binding type

high binding surface

Looking for similar products? Visit Product Comparison Guide

General description

MICROLON® 600, 96W 12xC8 Strip Microplate, working volume 20-300 μL; total well volume 346 μL.

Other Notes

96 well solid C-bottom microplate in white frame, C8 strips mounted in frame, high-binding, no lid, alphanumeric well coding, length: 127.76 mm; width: 85.48 mm.

Suitability

UsefμL in immunological assays. MICROLON 600 is a hydrophobic medium binding surface that will bind up to 600 ng/cm2 of non-polar proteins and peptides. Free of detectable DNase/RNase, human DNA and pyrogens.

Legal Information

MICROLON is a registered trademark of Greiner Bio-One GmbH

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service